pTracer™-CMV/Bsd Mammalian Expression Vector

Catalog number: V88320

Invitrogen™  Related applications: Cellular Imaging | Protein Assays and Analysis | Protein Expression

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Two Tracer™ mammalian expression vectors are available that express GFP fused to the selectable marker Blasticidin. Blasticidin is a potent selection agent that allows a more rapid generation of stable cell lines than other selection agents. Each vector uses a different set of promoters to express the gene of interest and the Cycle 3 GFP-Blasticidin resistance gene fusion.

The pTracer™-CMV/Bsd vector offers:

• CMV promoter for high-level constitutive expression of your gene of interest
• EF-1 promoter for high-level constitutive expression of the Cycle 3 GFP-Blasticidin resistance fusion

The pTracer™-EF/Bsd vectors offer:
• EF-1 promoter for high-level constitutive expression of your gene of interest
• CMV promoter for high-level constitutive expression of the Cycle 3 GFP-Blasticidin resistance fusion
• C-terminal V5 epitope tag for simple detection of recombinant protein with an Anti-V5 Antibody
• C-terminal 6xHis tag for rapid purification on nickel-chelating resin and simple detection with an Anti-His(C-term) Antibody
• Three reading frames for easier in-frame cloning
For Research Use Only. Not for use in diagnostic procedures.

Figures

Specifications

Promoter: CMV
Key Function: Reporter Assays
Protein Tag or Fusion: Untagged
Reporter: GFP (Cycle 3)
Delivery Method: Transfection
Cloning Method: Restriction Enzyme ⁄ MCS
Selection Agent (Eukaryotic): Blasticidin
Vector Type: pTracer
Constitutive or Inducible System: Constitutive
Product Line: pTRACER™
Product Size: 20 µg

Contents & storage

20 µg each of the pTracer™ /Bsd vector and a lacZ expression control are provided supercoiled and lyophilized. pTracer™ /Bsd Kits contain the appropriate pTracer™ vector and 50 mg of the selection agent Blasticidin. Store vectors and Blasticidin at -20°C. All reagents are stable for 6 months when properly stored.

Documents

Manuals & protocols

Recommended products