Related applications: Animal Health
Error loading your content!
Catalog number |
Select a plan
Unit size
|
Price ({{currency}}) | Availability | Qty | |
---|---|---|---|---|---|
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} |
|
— |
Target Organism Class: | Caprine Arthritis-Encephalitis Virus (CAEV), Visna-Maedi Virus |
---|---|
Validated Application: | ELISA |
Detection Method: | Colorimetric |
Sample Type (Specific): | Serum |
Product Line: | LSIVet™ |
Product Size: | 5 x 96 well plates |
Species: | Goat⁄Caprine, Sheep⁄Ovine |