Check out our new improved WesternDot® secondary antibodies in many species, sensitive, easier to use, multiplexable, and cheaper. Click here.

WesternDot® 625 Goat Anti-Rabbit Western Blot Kit

Catalog number: W10142

Molecular Probes™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

WesternDot® 625 Western Blot kits utilize extremely bright Qdot® 625 nanocrystals and include reagents for sensitive imaging of proteins probed with mouse or rabbit primary antibodies. With detection performance and ease-of-use that exceed existing ECL-based methods, the WesternDot® 625 kits make use of our extremely bright Qdot 625 streptavidin conjugate to allow direct imaging of blots while avoiding the need to spend time and money on optimizing multiple exposures to film in the dark. Each kit contains reagents sufficient for 20 minigel blots (7 cm × 10 cm/blot)
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reactivity: Rabbit
Color: Red
Label or Dye: Qdot® 625
Product Size: 1 kit
Detection Method: Fluorescent
Shipping Condition: Room Temperature
Sample Type (Specific): Cell Extracts, Protein
For Use With (Equipment): iBlot™, Photographic Imager

Contents & storage

WesternDot® blocking buffer (component A), Wash buffer (Component B), Biotin-XX goat anti-rabbit IgG (H+L) (Component C), Qdot® 625 streptavidin conjugate, (Component D), WesternDot® Staining dish (Component E), Store at 4°C, Do Not Freeze

Documents

Manuals & protocols

Recommended products