Invitrogen™

WesternBreeze™ Chromogenic Kit, anti-mouse

Catalog number:  WB7103

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

WesternBreeze® Chromogenic Kits detect proteins that have been immobilized on membranes (nitrocellulose or PVDF) following western transfer or bound directly from solution (dot blots). Detection is accomplished with a ready-to-use BCIP/NBT substrate for alkaline phosphatase. Non-fading purple precipitates develop at the protein bands on the membrane. WesternBreeze® chromogenic offers:

• Clear background
• High sensitivity-low picogram levels detectable
• High specificity
• One simple protocol-no optimization required
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Chromogenic
Product Line: WesternBreeze®
Product Size: 1 kit
Reactivity: Mouse
Substrate Type: AP (Alkaline Phosphatase) Substrate
Shipping Condition: Wet Ice

Contents & storage

The WesternBreeze® Chromogenic Kits include blocking solutions, primary antibody diluent, ready-to-use secondary antibody solution (anti-mouse, anti-rabbit or anti-goat), ready-to-use chromogenic substrate, wash solutions, and incubation trays. Each kit contains complete reagents for 20 mini-blots. Store the kits at +4°C. Guaranteed stable for 6 months when properly stored.

Documents

Manuals & protocols

Recommended products