Novex® AP Chromogenic Substrate (BCIP⁄ NBT)

Catalog number: WP20001

Novex™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Novex® AP Chromogenic Substrate is a ready to use single component reagent for sensitive immunodetection of alkaline phosphatase (AP) enzyme on western blot and dot blot membranes. Compatable for use with Western Breeze™ protocols, this ready-to-use solution of 5-bromo-4-chloro-3-indolyl-1-phosphate (BCIP) and nitroblue tetrazolium (NBT) will forms a black-purple precipitate upon reaction with alkaline phosphatase.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Colorimetric
Product Line: Novex™
Product Size: 250 mL
Substrate: BCIP⁄NBT
Substrate Properties: Chemical Substrate
Substrate Type: AP (Alkaline Phosphatase) Substrate
Shipping Condition: Approved for shipment at Room Temperature or on Wet Ice

Contents & storage

Single bottle labeled: Novex® AP Chromogenic Substrate (BCIP⁄NBT). Store at 4°C.

Documents

Manuals & protocols

Recommended products