Novex® AP Chemiluminescent Substrate (CDP-Star®)

Catalog number: WP20002

Novex™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Novex® AP Chemiluminescent Substrate is a non-radioactive ready-to-use solution of CDP-Star for chemiluminescence-based immunodetection of alkaline phosphatase (AP) on western blot or dot blot membranes. This reagent provides detection sensitivities superior to that of precipitating chromogenic substrates. Low picogram levels of detection can be achieved using either X-ray film or imaging equipment.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Chemiluminescent
Form: Liquid
Product Line: Novex™
Product Size: 100 mL
Substrate: CDP-Star®
Substrate Properties: Chemical Substrate
Substrate Type: AP (Alkaline Phosphatase) Substrate
Shipping Condition: Approved for shipment at Room Temperature or on Wet Ice

Contents & storage

Single bottle labeled: Novex® AP Chemiluminescent Substrate (CDP-Star®). Store at 4°C.

Documents

Manuals & protocols

Recommended products