Invitrogen™

Novex™ AP Chemiluminescent Substrate Enhancer (Nitro Block II™)

Catalog number:  WP20003

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Novex® AP Chemiluminescent Substrate Enhancer consists of a 20X solution of Nitro-Block-II™ intended for use in conjunction with Novex® AP Chemiluminescent Substrate to increase the intensity of chemiluminescent signals when probing blots on nitrocellulose membranes. The enhancer produces a hydrophobic environment on the membrane which increases signal intensity and allows shorter exposure times for signal acquisition.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Chemiluminescent
Form: Liquid
Product Line: Novex™
Product Size: 5 mL
Substrate: Nitro Block II™
Substrate Properties: Chemical Substrate
Substrate Type: AP (Alkaline Phosphatase) Substrate
Shipping Condition: Approved for shipment at Room Temperature or on Wet Ice

Contents & storage

Single bottle labeled: Novex® AP Chemiluminescent Substrate Enhancer (Nitro Block II™). Store at 4°C

Documents

Manuals & protocols

Recommended products