Invitrogen™

Novex™ HRP Chromogenic Substrate (TMB)

Catalog number:  WP20004

 Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Novex® HRP Chromogenic Substrate is a ready to use single component reagent used for sensitive detection of a horse radish peroxidase (HRP) on western blot and dot blot membranes. Consisting of a ready-to-use solution of TMB, the Novex® HRP Chromogenic Substrate forms a blue precipatate up reaction with horse radish peroxidase (HRP).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color: Blue
Detection Method: Colorimetric
Form: Liquid
Product Line: Novex™
Product Size: 250 mL
Quantity: 1 bottle
Substrate: TMB
Substrate Properties: Chemical Substrate
Substrate Type: HRP (Horseradish Peroxidase) Substrate
Shipping Condition: Approved for shipment at Room Temperature or on Wet Ice

Contents & storage

Single bottle labelled: Novex® HRP Chromogenic Substrate (TMB). Avoid exposure to light. Store at 4°C.

Documents

Manuals & protocols

Recommended products