The BenchPro® 4100 Card Processing Station will no longer be available for purchase after December 31, 2014. The current BenchPro® 4100 Western Cards and Reagent Vials will continue to be available for purchase. We are pleased to introduce the new iBind™ Western Device as an alternative system for automated western blot processing.

BenchPro™ 4100 Reagent Vials

Catalog number:  WP3001

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

BenchPro® 4100 Reagent Vials are available as a package of 50- 25ml vials. They are compatible for use with the BenchPro® 4100 Card Processing Station.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capacity: 25 mL
Compatible Products: Designed for use with WP0001
Dimensions: 50 tubes, 25 mL each
For Use With (Equipment): BenchPro® 4100 Western Processing System
Format: Tube(s)
Product Line: BenchPro™
Product Size: 50 vials
Shipping Condition: Room Temperature
Type: Vials

Contents & storage

50 graduated vials; Volume: 25mLs (each). Store at room temperature.

Documents

Manuals & protocols

Recommended products