Zenon® Alexa Fluor® 405 Mouse IgG2a Labeling Kit

Catalog number: Z25113

Invitrogen™  Related applications: Labeling Chemistry | Protein Labeling & Crosslinking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Zenon labeling technology provides a fast, versatile and reliable method for producing antibody conjugates, even with very small (submicrogram) amounts of starting material. Antibody conjugates formed using Zenon technology may be used to stain cells in any protocol where a directly labeled primary antibody is suitable, including flow cytometry, imaging, high throughput and other applications. Moreover, this technology simplifies applications that previously were time consuming or not practical, such as the use of multiple mouse-derived antibodies in the same staining protocol.

View a selection guide for all Zenon™ antibody labeling kits and other antibody labeling products.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Labeling Method: Zenon® Secondary Detection-Based
Detection Method: Fluorescent
Excitation⁄Emission (nm): 401⁄421
Flow Cytometer Laser Lines: 405
Label or Dye: Alexa Fluor™ 405
Labeling Scale: < 1-20 µg
Labeling Target: Antibodies (General)
Number of Reactions: 50
Product Line: Alexa Fluor®, ZENON®
Product Size: 1 kit
Reactivity: Mouse
Shipping Condition: Room Temperature
Target Isotype: IgG2a

Contents & storage

Contains 1 vial of Zenon® Alexa Fluor® 405 mouse IgG2a labeling reagent (250 µL), and 1 vial of Zenon® blocking reagent (mouse IgG, 250 µL).

Store in refrigerator (2–8°C) and protect from light.

Documents

Manuals & protocols

Recommended products