ZOOM™ Equilibration Trays

Catalog number:  ZM0007

 Related applications: Protein Gel Electrophoresis | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

ZOOM® Equilibration Trays are convenient, disposable trays designed for use with the ZOOM® IPGRunner™ Cassettes to perform equilibration of ZOOM® Strips after first-dimension isoelectric focusing and prior to second-dimension (2D) SDS-PAGE (Figure 1).

The ZOOM® Equilibration Trays simplify the equilibration and incubation process in 2D electrophoresis and offer following advantages:

• Savings in time and labor
• Optimal equilibration of samples
• Minimal risk of sample contamination and damage to IPG strips
For Research Use Only. Not for use in diagnostic procedures.

Figures

Specifications

For Use With (Equipment): ZOOM® IPGRunner™ Mini
Product Line: ZOOM®
Product Size: 10⁄pack
Type: Tray
Shipping Condition: Room Temperature

Contents & storage

The ZOOM® Equilibration Trays are supplied in a pack of 10. The combo kit of ZOOM®Cassettes and ZOOM® Equilibration Trays includes a box of 10 cassettes and a pack of 10 trays. Store at room temperature. Guaranteed stable for one year when properly stored.

Documents

Manuals & protocols

Recommended products