ZOOM® Carrier Ampholytes pH 3-10

Catalog number: ZM0021

Invitrogen™  Related applications: Protein Gel Electrophoresis | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The ZOOM® IPGRunner™ System is the first system to offer oil-free, trouble-free 2D electrophoresis in a mini-gel format. First dimension separation, isoelectric focusing (IEF), is complete in less than three hours. The mini-gel design of the system is easy to handle, eliminates mineral oil overlays, and allows processing of up to 12 samples at once for high-throughput usage. The ZOOM® IPGRunner™ System consists of three main components:

• The ZOOM® IPGRunner™ Mini-Cell (Figure 1) includes a high-voltage electrophoresis core and lid assembly that fits a mini-cell chamber identical to the XCell SureLock™ mini-cell. The mini-cell holds two ZOOM® IPGRunner™ Cassettes.
• The ZOOM® IPGRunner™ Cassettes (Figure 2) are provided ready to use and self-contained, eliminating oil overlays for convenient sample rehydration and IEF. Each cassette holds up to six ZOOM® Strips.
• The ZOOM® Strips are 7 cm long and contain a thin layer of polyacrylamide gel that includes a fixed pH gradient. Each ZOOM® Strip is clearly labeled with a unique identifying number, pH range, and orientation marks (Figure 3). ZOOM® Strips are supplied attached to a tri-fold card (Figure 4) for easy access and removal.

Quick and Simple Set-up Procedure
Figure 5 demonstrates how easy and fast it is to load samples and set up the ZOOM® IPGRunner™ System for IEF. Once IEF is complete, the ZOOM® IPGRunner™ Mini-Cell can be used with NuPAGE® and Novex® ZOOM® gels to complete SDS-PAGE analysis in 40 minutes.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line: ZOOM®
Product Size: 10 mL
Shipping Condition: Room Temperature

Contents & storage

The ZOOM® IPGRunner™ Mini-Cell includes the ZOOM® IPGRunner™ Buffer Core and Lid, Gel Tension Wedge, Buffer Dam, and the Mini- Cell chamber. The ZOOM® IPGRunner™ Combo Kit includes the ZOOM® IPGRunner™ Mini-Cell, electrode wicks, sealing tape, 10 ZOOM® IPGRunner™ Cassettes, and 12 ZOOM® Strips, pH 3-10 NL. The ZOOM® IPGRunner® Retrofit Kit for XCell SureLock™ Mini-Cell includes the ZOOM® IPGRunner™ Buffer Core and Lid. The ZOOM® IPGRunner™ Cassettes include electrode wicks and sealing tape.

Documents

Manuals & protocols

Recommended products