ZOOM® Dual Power Supply (100-120 VAC 47 - 60 Hz) œ

Catalog number: ZP10001

 Related applications: Protein Assays and Analysis | Protein Gel Electrophoresis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The ZOOM® Dual Power is a microprocessor-controlled programmable power supply capable of running both high voltage/low current and high current/low voltage applications concurrently. The high voltage/low current section is ideal for sample fractionation, IPG strips, IEF, and DNA sequencing applications. The low voltage/high current section is for SDS-PAGE, native PAGE, second-dimension SDS-PAGE, DNA/RNA electrophoresis, and blotting applications. Four sets of output jacks on each side of the unit allow for multiple apparatus to run simultaneously, offering greater workflow efficiency and higher sample throughput. Some of the important features of the ZOOM® Dual Power unit include:
• High voltage/low current and low voltage/high current sides can be used independently and concurrently
• Multiple output jacks allow for the simultaneous running of up to four electrophoresis apparatus on each side of the unit
• Memory stores up to 30 programs (15 programs per side) of up to six steps per program. Step and ramp functions for discrete and gradual changes of electrical parameters
• No Load detection alarm system can be disabled to power high voltage applications that require very low current

Specifications of the ZOOM® Dual Power
High voltage/Low current output range




Voltage: 1–3500 V; adjustable in 1V steps
Current: 0.01–10.0 mA, adjustable in 0.01 mA increments
Power: 0.1– 35W in 0.1 W steps

Low voltage/High current



output range


Voltage: 1–250 V; adjustable in 1V steps
Current: 1–800 mA, adjustable in 1 mA increments
Power: 1–200 W in 1 W steps

Power Outlets

4 pairs of terminals

Programs Stored

15 programs, up to 6 steps each

Duration Limits

Time: 1 to 9999 min in 1 min steps
Voltage Hours

:

1 to 65000 V-hr in 1 V-hr steps

Operating Modes

Constant voltage, constant current, constant power

Circuit Protection

Open Circuit; Short Circuit; Thermal Protection Over Voltage, Over Current, Over Power

Safety

No Load Detection Enable/Disable Isolated Floating Ground

Memory Control

Non-volatile, up to 24 hours. Custom Methods Stored to RAM

Display

LCD, view area 111.7 x 86mm

Temperature

Ambient 4-40°C

Dimensions

23.5 cm (w) x 34.5 cm (h) x 31 cm (d) (37 cm (d) with power cord attached

Weight

12 kg (26.5 lbs)

Applications for the ZOOM® Dual Power


High voltage/low current


Suggested Applications

• Isoelectric focusing in tube gels
• Isoelectric focusing in IPG strips
• Solution phase isoelectric focusing

Electrophoresis Apparatus:

• ZOOM® IPGRunner™ system
• ZOOM® IEF Fractionators

Low voltage/High current
Suggested Applications

• Blotting applications
• DNA and RNA electrophoresis
• SDS-PAGE
• Native PAGE

Electrophoresis Apparatus:

• XCell SureLock® Mini-Cell
• XCell4 SureLock® Midi-Cell
• XCell6™ Multigel Unit
• XCell II™ Blot Module
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): ZOOM® Dual Power
Product Line: ZOOM®
Product Size: 1 each
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols

Recommended products