DQ™ Red BSA  - Special Packaging

Catalog number: D12051

Invitrogen™  Related applications: Cell Viability, Proliferation & Function | Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

DQ Red BSA is a fluorogenic substrate for proteases. A strong fluorescence quenching effect is observed when proteins are heavily labeled with BODIPY dyes. Upon hydrolysis of the DQ Red BSA to single, dye-labeled peptides by proteases, this quenching is relieved, producing brightly fluorescent products.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Fluorescent
Product Line: DQ™
Product Size: 5 x 1 mg
Substrate: DQ™ Red BSA
Substrate Properties: Protein-Based Substrate
Substrate Type: Protease Substrate
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C) and protect from light.

Documents

Manuals & protocols

Recommended products