CD39 / ENTPD1 Antibody (A1), Molecular Probes™

Catalog number: MHCD3900

Invitrogen™  Related applications: Flow Cytometry

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CD39 cell surface antigen is a 70-100kD molecule expressed on peripheral blood B cells, monocytes, T cell clones, and also weakly expressed on granulocytes. CD39 has intrinsic ecto-ATPase activity. Expression of CD39 is induced on T cells and increased on B cells as a late activation antigen.
Analyte Specific Reagent. Analytical and performance characteristics are not established.

Specifications

Reactivity: Human
Validated Application: Flow Cytometry
Host Species: Mouse
Clonality: Monoclonal
Clone ID: A1
Target Gene: CD39
Gene ID: 953
Label or Dye: Unconjugated
Number of Reactions: 100 Tests
Product Size: 0.5 ml
Shipping Condition: Wet Ice

Contents & storage

Store reagents at 2-8°C.

Documents

Manuals & protocols

Recommended products