Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 39136875-39137374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GEMIN2
Entrez Gene ID
8487
Gene Name
gem nuclear organelle associated protein 2
Gene Aliases
SIP1, SIP1-delta
UniGene
Hs.652307
Gene Chromosome Location
Chr.14: 39114261-39136973 on Build GRCh38
Transcripts
NM_001009182.1 NM_001009183.1 NM_003616.2 XM_017021709.1

SNP Detail

SNP ID Chromosome Location
rs1020024338 Chr.14: 39137022-39137022 on Build GRCh38
rs776367441 Chr.14: 39137109-39137122 on Build GRCh38
rs1035382777 Chr.14: 39137068-39137068 on Build GRCh38
rs576194425 Chr.14: 39137188-39137188 on Build GRCh38
rs12586309 Chr.14: 39136999-39136999 on Build GRCh38
rs866072315 Chr.14: 39136998-39136998 on Build GRCh38
rs942714433 Chr.14: 39137239-39137239 on Build GRCh38
rs562534226 Chr.14: 39137141-39137141 on Build GRCh38
rs892946958 Chr.14: 39137009-39137009 on Build GRCh38
rs912535081 Chr.14: 39137235-39137235 on Build GRCh38
rs10130011 Chr.14: 39137002-39137002 on Build GRCh38
rs747012141 Chr.14: 39136983-39136984 on Build GRCh38
rs1026836594 Chr.14: 39137222-39137222 on Build GRCh38
rs948461664 Chr.14: 39136938-39136938 on Build GRCh38
rs962879287 Chr.14: 39137145-39137145 on Build GRCh38
rs938577091 Chr.14: 39137258-39137258 on Build GRCh38
rs974683349 Chr.14: 39137149-39137149 on Build GRCh38
rs1006838894 Chr.14: 39136915-39136915 on Build GRCh38
rs958370953 Chr.14: 39137092-39137092 on Build GRCh38
rs564784144 Chr.14: 39137215-39137215 on Build GRCh38
rs574160655 Chr.14: 39137043-39137043 on Build GRCh38
rs1001128286 Chr.14: 39137052-39137052 on Build GRCh38
rs903997817 Chr.14: 39137024-39137024 on Build GRCh38
rs908492631 Chr.14: 39137257-39137257 on Build GRCh38
rs1002520387 Chr.14: 39137026-39137026 on Build GRCh38
rs1039589973 Chr.14: 39137295-39137295 on Build GRCh38
rs201921488 Chr.14: 39137011-39137011 on Build GRCh38
rs1045249616 Chr.14: 39137006-39137006 on Build GRCh38
rs918670759 Chr.14: 39137172-39137172 on Build GRCh38
rs868084670 Chr.14: 39137117-39137117 on Build GRCh38
rs796321738 Chr.14: 39137309-39137309 on Build GRCh38
rs761648244 Chr.14: 39137244-39137244 on Build GRCh38
rs767802152 Chr.14: 39136984-39136984 on Build GRCh38
rs998393152 Chr.14: 39136981-39136981 on Build GRCh38
rs567890648 Chr.14: 39137000-39137005 on Build GRCh38
rs75039627 Chr.14: 39137000-39137000 on Build GRCh38
rs142110324 Chr.14: 39137328-39137328 on Build GRCh38
rs554244152 Chr.14: 39136912-39136912 on Build GRCh38
rs57533780 Chr.14: 39137001-39137001 on Build GRCh38
rs544927637 Chr.14: 39137238-39137238 on Build GRCh38
rs375590121 Chr.14: 39137140-39137140 on Build GRCh38
rs61515013 Chr.14: 39136996-39136996 on Build GRCh38
rs930086866 Chr.14: 39137180-39137180 on Build GRCh38
rs774612050 Chr.14: 39136982-39136982 on Build GRCh38
rs796864341 Chr.14: 39136982-39136983 on Build GRCh38
rs751235847 Chr.14: 39136982-39136983 on Build GRCh38
rs66675441 Chr.14: 39136983-39136983 on Build GRCh38
rs967158557 Chr.14: 39137023-39137023 on Build GRCh38

View Full Product Details