Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 75633535-75634078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLVCR2
Entrez Gene ID
55640
Gene Name
feline leukemia virus subgroup C cellular receptor family member 2
Gene Aliases
C14orf58, CCT, EPV, FLVCRL14q, MFSD7C, PVHH
UniGene
Hs.509966
Gene Chromosome Location
Chr.14: 75578597-75648169 on Build GRCh38
Transcripts
NM_001195283.1 NM_017791.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6554529 c.1013A>T p.H338L Chr.14: 75633689-75633689 on Build GRCh38
363260 c.1005G>T p.V335V Chr.14: 75633681-75633681 on Build GRCh38
5508592 c.953-6delC p.? Chr.14: 75633623-75633623 on Build GRCh38
6449275 c.1020G>A p.P340P Chr.14: 75633696-75633696 on Build GRCh38
957996 c.998G>A p.R333H Chr.14: 75633674-75633674 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951641411 Chr.14: 75633894-75633894 on Build GRCh38
rs890961359 Chr.14: 75633711-75633711 on Build GRCh38
rs763074367 Chr.14: 75633662-75633662 on Build GRCh38
rs370436338 Chr.14: 75633579-75633579 on Build GRCh38
rs866932069 Chr.14: 75633850-75633850 on Build GRCh38
rs562124005 Chr.14: 75633789-75633789 on Build GRCh38
rs150334666 Chr.14: 75633560-75633560 on Build GRCh38
rs995160689 Chr.14: 75633808-75633808 on Build GRCh38
rs897115944 Chr.14: 75633929-75633929 on Build GRCh38
rs1026400702 Chr.14: 75633618-75633618 on Build GRCh38
rs759469907 Chr.14: 75633686-75633686 on Build GRCh38
rs778576250 Chr.14: 75633818-75633818 on Build GRCh38
rs1038656904 Chr.14: 75633923-75633923 on Build GRCh38
rs759352677 Chr.14: 75633739-75633739 on Build GRCh38
rs769534566 Chr.14: 75633745-75633745 on Build GRCh38
rs1017695736 Chr.14: 75634036-75634036 on Build GRCh38
rs928786433 Chr.14: 75634009-75634009 on Build GRCh38
rs763033639 Chr.14: 75633601-75633601 on Build GRCh38
rs756480123 Chr.14: 75633696-75633696 on Build GRCh38
rs267606824 Chr.14: 75633653-75633653 on Build GRCh38
rs772859417 Chr.14: 75634015-75634015 on Build GRCh38
rs768301565 Chr.14: 75633605-75633605 on Build GRCh38
rs555834563 Chr.14: 75633893-75633893 on Build GRCh38
rs1009366582 Chr.14: 75634016-75634016 on Build GRCh38
rs111529038 Chr.14: 75633676-75633676 on Build GRCh38
rs115200097 Chr.14: 75633615-75633615 on Build GRCh38
rs1050918092 Chr.14: 75633694-75633694 on Build GRCh38
rs189794315 Chr.14: 75633889-75633889 on Build GRCh38
rs750773606 Chr.14: 75633695-75633695 on Build GRCh38
rs375448021 Chr.14: 75633713-75633713 on Build GRCh38
rs34788319 Chr.14: 75633628-75633628 on Build GRCh38
rs776582879 Chr.14: 75633729-75633729 on Build GRCh38
rs529450422 Chr.14: 75633979-75633979 on Build GRCh38
rs949863314 Chr.14: 75633805-75633805 on Build GRCh38
rs188707531 Chr.14: 75633614-75633614 on Build GRCh38
rs535164675 Chr.14: 75634020-75634020 on Build GRCh38
rs533371902 Chr.14: 75633774-75633774 on Build GRCh38
rs184888851 Chr.14: 75633775-75633775 on Build GRCh38
rs747667067 Chr.14: 75633607-75633607 on Build GRCh38
rs781633360 Chr.14: 75633603-75633603 on Build GRCh38
rs752168189 Chr.14: 75633583-75633583 on Build GRCh38
rs774571869 Chr.14: 75633609-75633609 on Build GRCh38
rs991657715 Chr.14: 75634011-75634011 on Build GRCh38
rs755604627 Chr.14: 75633678-75633678 on Build GRCh38
rs1042139499 Chr.14: 75633639-75633639 on Build GRCh38
rs766648145 Chr.14: 75633599-75633601 on Build GRCh38
rs114938064 Chr.14: 75633790-75633790 on Build GRCh38
rs199607525 Chr.14: 75633606-75633606 on Build GRCh38
rs760675970 Chr.14: 75633616-75633616 on Build GRCh38
rs746245889 Chr.14: 75633604-75633604 on Build GRCh38
rs765459389 Chr.14: 75633656-75633656 on Build GRCh38
rs181232141 Chr.14: 75633620-75633620 on Build GRCh38
rs982114954 Chr.14: 75634004-75634004 on Build GRCh38
rs778610642 Chr.14: 75633702-75633702 on Build GRCh38
rs372118257 Chr.14: 75633624-75633624 on Build GRCh38
rs114384838 Chr.14: 75633770-75633770 on Build GRCh38
rs771542306 Chr.14: 75634045-75634045 on Build GRCh38
rs1025756931 Chr.14: 75633814-75633814 on Build GRCh38
rs374886567 Chr.14: 75633581-75633581 on Build GRCh38
rs374709682 Chr.14: 75633619-75633619 on Build GRCh38
rs930251002 Chr.14: 75633563-75633563 on Build GRCh38
rs200712122 Chr.14: 75633717-75633717 on Build GRCh38
rs745518486 Chr.14: 75634010-75634010 on Build GRCh38
rs138853227 Chr.14: 75633658-75633658 on Build GRCh38
rs747816671 Chr.14: 75633705-75633705 on Build GRCh38
rs770807093 Chr.14: 75633718-75633718 on Build GRCh38
rs757778790 Chr.14: 75633674-75633674 on Build GRCh38
rs764144834 Chr.14: 75633673-75633673 on Build GRCh38
rs757690678 Chr.14: 75633594-75633594 on Build GRCh38

View Full Product Details