Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50655490-50655995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC3
Entrez Gene ID
8714
Gene Name
ATP binding cassette subfamily C member 3
Gene Aliases
ABC31, EST90757, MLP2, MOAT-D, MRP3, cMOAT2
UniGene
Hs.463421
Gene Chromosome Location
Chr.17: 50634843-50691704 on Build GRCh38
Transcripts
NM_001144070.1 NM_003786.3 XM_005257763.3 XM_011525422.2 XM_011525423.1 XM_011525425.2 XM_017025266.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5551252 c.153C>G p.C51W Chr.17: 50655939-50655939 on Build GRCh38
5706457 c.77C>T p.P26L Chr.17: 50655863-50655863 on Build GRCh38
5933517 c.89C>T p.P30L Chr.17: 50655875-50655875 on Build GRCh38
5004361 c.120G>A p.V40V Chr.17: 50655906-50655906 on Build GRCh38
4396896 c.79G>A p.D27N Chr.17: 50655865-50655865 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1007957151 Chr.17: 50655711-50655711 on Build GRCh38
rs764673801 Chr.17: 50655782-50655782 on Build GRCh38
rs201851809 Chr.17: 50655921-50655921 on Build GRCh38
rs896676321 Chr.17: 50655656-50655656 on Build GRCh38
rs532892311 Chr.17: 50655555-50655555 on Build GRCh38
rs749910845 Chr.17: 50655784-50655784 on Build GRCh38
rs564196865 Chr.17: 50655536-50655536 on Build GRCh38
rs928470445 Chr.17: 50655641-50655641 on Build GRCh38
rs376420023 Chr.17: 50655797-50655797 on Build GRCh38
rs1032915256 Chr.17: 50655752-50655752 on Build GRCh38
rs200070901 Chr.17: 50655955-50655955 on Build GRCh38
rs762519135 Chr.17: 50655790-50655790 on Build GRCh38
rs908336862 Chr.17: 50655847-50655847 on Build GRCh38
rs964276059 Chr.17: 50655756-50655756 on Build GRCh38
rs550248080 Chr.17: 50655687-50655687 on Build GRCh38
rs768886195 Chr.17: 50655952-50655952 on Build GRCh38
rs941056615 Chr.17: 50655550-50655550 on Build GRCh38
rs767241920 Chr.17: 50655965-50655965 on Build GRCh38
rs373210147 Chr.17: 50655794-50655794 on Build GRCh38
rs531113862 Chr.17: 50655530-50655530 on Build GRCh38
rs149367152 Chr.17: 50655839-50655839 on Build GRCh38
rs780977211 Chr.17: 50655799-50655799 on Build GRCh38
rs62059746 Chr.17: 50655826-50655826 on Build GRCh38
rs948778844 Chr.17: 50655606-50655606 on Build GRCh38
rs373449095 Chr.17: 50655861-50655861 on Build GRCh38
rs141743169 Chr.17: 50655645-50655645 on Build GRCh38
rs751264707 Chr.17: 50655874-50655874 on Build GRCh38
rs759146021 Chr.17: 50655959-50655959 on Build GRCh38
rs1009617459 Chr.17: 50655677-50655677 on Build GRCh38
rs762606673 Chr.17: 50655868-50655868 on Build GRCh38
rs955214541 Chr.17: 50655804-50655804 on Build GRCh38
rs902004470 Chr.17: 50655744-50655744 on Build GRCh38
rs544850618 Chr.17: 50655534-50655534 on Build GRCh38
rs756031850 Chr.17: 50655825-50655825 on Build GRCh38
rs866383362 Chr.17: 50655887-50655887 on Build GRCh38
rs746023660 Chr.17: 50655854-50655854 on Build GRCh38
rs531734645 Chr.17: 50655652-50655652 on Build GRCh38
rs140077054 Chr.17: 50655864-50655864 on Build GRCh38
rs774985556 Chr.17: 50655967-50655967 on Build GRCh38
rs984718700 Chr.17: 50655554-50655554 on Build GRCh38
rs770034967 Chr.17: 50655905-50655906 on Build GRCh38
rs184202680 Chr.17: 50655518-50655518 on Build GRCh38
rs767240602 Chr.17: 50655886-50655886 on Build GRCh38
rs145596140 Chr.17: 50655927-50655927 on Build GRCh38
rs772296089 Chr.17: 50655931-50655931 on Build GRCh38
rs147484871 Chr.17: 50655848-50655848 on Build GRCh38
rs780105010 Chr.17: 50655938-50655938 on Build GRCh38
rs775874639 Chr.17: 50655863-50655863 on Build GRCh38
rs370292346 Chr.17: 50655569-50655569 on Build GRCh38
rs769243628 Chr.17: 50655865-50655865 on Build GRCh38
rs553304862 Chr.17: 50655956-50655956 on Build GRCh38
rs975373756 Chr.17: 50655770-50655770 on Build GRCh38
rs1001313749 Chr.17: 50655698-50655698 on Build GRCh38
rs748218315 Chr.17: 50655821-50655822 on Build GRCh38
rs753786386 Chr.17: 50655971-50655971 on Build GRCh38
rs188529531 Chr.17: 50655651-50655651 on Build GRCh38
rs757334041 Chr.17: 50655928-50655928 on Build GRCh38
rs542239034 Chr.17: 50655516-50655516 on Build GRCh38
rs745495349 Chr.17: 50655962-50655962 on Build GRCh38
rs752593528 Chr.17: 50655801-50655801 on Build GRCh38
rs1046252944 Chr.17: 50655615-50655615 on Build GRCh38
rs751239962 Chr.17: 50655796-50655796 on Build GRCh38
rs146229726 Chr.17: 50655678-50655678 on Build GRCh38
rs370621836 Chr.17: 50655689-50655689 on Build GRCh38
rs548849536 Chr.17: 50655712-50655712 on Build GRCh38
rs777252908 Chr.17: 50655867-50655867 on Build GRCh38
rs1038612516 Chr.17: 50655591-50655591 on Build GRCh38
rs763280136 Chr.17: 50655727-50655727 on Build GRCh38
rs772336608 Chr.17: 50655855-50655855 on Build GRCh38
rs376156397 Chr.17: 50655930-50655930 on Build GRCh38
rs889472637 Chr.17: 50655710-50655710 on Build GRCh38
rs752498333 Chr.17: 50655894-50655894 on Build GRCh38
rs765978466 Chr.17: 50655872-50655872 on Build GRCh38
rs755835594 Chr.17: 50655916-50655916 on Build GRCh38
rs763434974 Chr.17: 50655866-50655866 on Build GRCh38
rs371251448 Chr.17: 50655946-50655946 on Build GRCh38
rs915920827 Chr.17: 50655600-50655600 on Build GRCh38
rs759174103 Chr.17: 50655875-50655875 on Build GRCh38
rs760576754 Chr.17: 50655968-50655968 on Build GRCh38

View Full Product Details