Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 13883794-13884290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MC2R
Entrez Gene ID
4158
Gene Name
melanocortin 2 receptor
Gene Aliases
ACTHR
UniGene
Hs.248144
Gene Chromosome Location
Chr.18: 13882042-13915707 on Build GRCh38
Transcripts
NM_000529.2 NM_001291911.1 XM_017025781.1

SNP Detail

SNP ID Chromosome Location
rs1020466478 Chr.18: 13884246-13884246 on Build GRCh38
rs34861769 Chr.18: 13884043-13884043 on Build GRCh38
rs545401448 Chr.18: 13884214-13884214 on Build GRCh38
rs556477756 Chr.18: 13883969-13883969 on Build GRCh38
rs558924106 Chr.18: 13884265-13884265 on Build GRCh38
rs139176501 Chr.18: 13884090-13884090 on Build GRCh38
rs555981788 Chr.18: 13884000-13884000 on Build GRCh38
rs754283731 Chr.18: 13883834-13883834 on Build GRCh38
rs749097541 Chr.18: 13884235-13884235 on Build GRCh38
rs371971294 Chr.18: 13884028-13884028 on Build GRCh38
rs1044875778 Chr.18: 13883949-13883949 on Build GRCh38
rs750654248 Chr.18: 13883844-13883844 on Build GRCh38
rs970564983 Chr.18: 13884014-13884014 on Build GRCh38
rs561276395 Chr.18: 13884086-13884086 on Build GRCh38
rs1052892077 Chr.18: 13883996-13883996 on Build GRCh38
rs56150730 Chr.18: 13884076-13884076 on Build GRCh38
rs1040980097 Chr.18: 13884239-13884239 on Build GRCh38
rs753026395 Chr.18: 13883951-13883951 on Build GRCh38
rs34482956 Chr.18: 13884047-13884047 on Build GRCh38
rs1035693532 Chr.18: 13884215-13884215 on Build GRCh38
rs538470189 Chr.18: 13883958-13883958 on Build GRCh38
rs886053647 Chr.18: 13884191-13884191 on Build GRCh38
rs755326782 Chr.18: 13883916-13883916 on Build GRCh38
rs373694705 Chr.18: 13884153-13884153 on Build GRCh38
rs543632552 Chr.18: 13884091-13884091 on Build GRCh38
rs777765132 Chr.18: 13884040-13884040 on Build GRCh38
rs943117988 Chr.18: 13884123-13884123 on Build GRCh38
rs951145782 Chr.18: 13883859-13883859 on Build GRCh38
rs969102027 Chr.18: 13884143-13884143 on Build GRCh38
rs768500810 Chr.18: 13884259-13884259 on Build GRCh38
rs1006296210 Chr.18: 13883989-13883989 on Build GRCh38
rs1016106640 Chr.18: 13884190-13884190 on Build GRCh38
rs1025771784 Chr.18: 13884121-13884121 on Build GRCh38
rs1007632731 Chr.18: 13884238-13884238 on Build GRCh38
rs578163818 Chr.18: 13883977-13883977 on Build GRCh38
rs980087448 Chr.18: 13884042-13884042 on Build GRCh38
rs1026147814 Chr.18: 13883973-13883973 on Build GRCh38
rs951030864 Chr.18: 13884131-13884131 on Build GRCh38
rs28926183 Chr.18: 13884134-13884134 on Build GRCh38
rs4517858 Chr.18: 13884018-13884018 on Build GRCh38
rs1052357564 Chr.18: 13884196-13884196 on Build GRCh38
rs142527936 Chr.18: 13884154-13884154 on Build GRCh38
rs997874514 Chr.18: 13883964-13883964 on Build GRCh38
rs886053648 Chr.18: 13884224-13884224 on Build GRCh38
rs886053645 Chr.18: 13884017-13884017 on Build GRCh38
rs553257738 Chr.18: 13884219-13884219 on Build GRCh38
rs35749889 Chr.18: 13884057-13884058 on Build GRCh38
rs1006034489 Chr.18: 13884170-13884170 on Build GRCh38
rs35053989 Chr.18: 13884109-13884109 on Build GRCh38
rs532094531 Chr.18: 13884136-13884136 on Build GRCh38
rs4308014 Chr.18: 13883940-13883940 on Build GRCh38
rs903517775 Chr.18: 13883913-13883913 on Build GRCh38
rs150928586 Chr.18: 13884198-13884198 on Build GRCh38
rs1039130161 Chr.18: 13883952-13883952 on Build GRCh38
rs1043790812 Chr.18: 13884045-13884045 on Build GRCh38
rs886367848 Chr.18: 13884157-13884157 on Build GRCh38
rs886053649 Chr.18: 13884249-13884249 on Build GRCh38
rs925035330 Chr.18: 13884101-13884101 on Build GRCh38
rs886053646 Chr.18: 13884155-13884155 on Build GRCh38
rs978914291 Chr.18: 13884095-13884095 on Build GRCh38
rs4464147 Chr.18: 13884010-13884010 on Build GRCh38
rs1007702524 Chr.18: 13884071-13884071 on Build GRCh38
rs950552887 Chr.18: 13883983-13883983 on Build GRCh38

View Full Product Details