Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41194302-41194707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2S1
Entrez Gene ID
29785
Gene Name
cytochrome P450 family 2 subfamily S member 1
Gene Aliases
CYPIIS1
UniGene
Hs.98370
Gene Chromosome Location
Chr.19: 41193207-41207539 on Build GRCh38
Transcripts
NM_030622.7

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7101890 c.334G>C p.D112H Chr.19: 41194700-41194700 on Build GRCh38
5989587 c.182G>T p.S61I Chr.19: 41194548-41194548 on Build GRCh38
4877597 c.192C>T p.Y64Y Chr.19: 41194558-41194558 on Build GRCh38
1712333 c.308C>T p.T103I Chr.19: 41194674-41194674 on Build GRCh38
4078421 c.229C>T p.P77S Chr.19: 41194595-41194595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567399913 Chr.19: 41194432-41194432 on Build GRCh38
rs376137528 Chr.19: 41194629-41194629 on Build GRCh38
rs201188229 Chr.19: 41194681-41194681 on Build GRCh38
rs900020910 Chr.19: 41194349-41194349 on Build GRCh38
rs763116277 Chr.19: 41194560-41194560 on Build GRCh38
rs117216419 Chr.19: 41194441-41194441 on Build GRCh38
rs200179278 Chr.19: 41194479-41194479 on Build GRCh38
rs202100446 Chr.19: 41194565-41194565 on Build GRCh38
rs141962525 Chr.19: 41194676-41194676 on Build GRCh38
rs776619879 Chr.19: 41194538-41194538 on Build GRCh38
rs763179944 Chr.19: 41194634-41194634 on Build GRCh38
rs60694775 Chr.19: 41194564-41194564 on Build GRCh38
rs933254997 Chr.19: 41194442-41194442 on Build GRCh38
rs926100896 Chr.19: 41194601-41194601 on Build GRCh38
rs768131385 Chr.19: 41194568-41194568 on Build GRCh38
rs956359151 Chr.19: 41194418-41194418 on Build GRCh38
rs373910816 Chr.19: 41194675-41194675 on Build GRCh38
rs993988588 Chr.19: 41194515-41194515 on Build GRCh38
rs773395536 Chr.19: 41194559-41194559 on Build GRCh38
rs199728850 Chr.19: 41194664-41194664 on Build GRCh38
rs764499716 Chr.19: 41194670-41194670 on Build GRCh38
rs910491067 Chr.19: 41194684-41194684 on Build GRCh38
rs370561000 Chr.19: 41194663-41194663 on Build GRCh38
rs75274897 Chr.19: 41194330-41194330 on Build GRCh38
rs746499193 Chr.19: 41194608-41194608 on Build GRCh38
rs1000605392 Chr.19: 41194360-41194360 on Build GRCh38
rs200051547 Chr.19: 41194548-41194548 on Build GRCh38
rs374503413 Chr.19: 41194628-41194628 on Build GRCh38
rs555319320 Chr.19: 41194640-41194640 on Build GRCh38
rs753314095 Chr.19: 41194573-41194573 on Build GRCh38
rs981115731 Chr.19: 41194453-41194453 on Build GRCh38
rs1032131040 Chr.19: 41194395-41194395 on Build GRCh38
rs773012943 Chr.19: 41194631-41194631 on Build GRCh38
rs771065124 Chr.19: 41194647-41194647 on Build GRCh38
rs527326617 Chr.19: 41194356-41194356 on Build GRCh38
rs338599 Chr.19: 41194588-41194588 on Build GRCh38
rs781628046 Chr.19: 41194439-41194439 on Build GRCh38
rs771576619 Chr.19: 41194499-41194499 on Build GRCh38
rs758006222 Chr.19: 41194593-41194593 on Build GRCh38
rs372840068 Chr.19: 41194454-41194454 on Build GRCh38
rs376296553 Chr.19: 41194513-41194513 on Build GRCh38
rs746913039 Chr.19: 41194534-41194534 on Build GRCh38
rs769539360 Chr.19: 41194630-41194630 on Build GRCh38
rs202034225 Chr.19: 41194680-41194680 on Build GRCh38
rs754457887 Chr.19: 41194679-41194679 on Build GRCh38
rs371117472 Chr.19: 41194625-41194625 on Build GRCh38
rs338600 Chr.19: 41194438-41194438 on Build GRCh38
rs369539924 Chr.19: 41194539-41194539 on Build GRCh38
rs150250299 Chr.19: 41194667-41194667 on Build GRCh38
rs779555227 Chr.19: 41194606-41194606 on Build GRCh38
rs978954970 Chr.19: 41194595-41194595 on Build GRCh38
rs754164611 Chr.19: 41194672-41194672 on Build GRCh38
rs756997221 Chr.19: 41194477-41194477 on Build GRCh38
rs1037477404 Chr.19: 41194481-41194481 on Build GRCh38
rs778318933 Chr.19: 41194586-41194586 on Build GRCh38
rs745564289 Chr.19: 41194496-41194496 on Build GRCh38
rs138879300 Chr.19: 41194668-41194668 on Build GRCh38
rs548986663 Chr.19: 41194423-41194423 on Build GRCh38
rs1026318802 Chr.19: 41194533-41194533 on Build GRCh38
rs201821032 Chr.19: 41194665-41194665 on Build GRCh38
rs201303420 Chr.19: 41194492-41194492 on Build GRCh38
rs540126374 Chr.19: 41194537-41194537 on Build GRCh38
rs756524311 Chr.19: 41194574-41194574 on Build GRCh38
rs925049157 Chr.19: 41194350-41194350 on Build GRCh38
rs888959666 Chr.19: 41194475-41194475 on Build GRCh38
rs865826120 Chr.19: 41194641-41194641 on Build GRCh38
rs62119652 Chr.19: 41194563-41194563 on Build GRCh38

View Full Product Details