Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 31924382-31924880 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HUNK
Entrez Gene ID
30811
Gene Name
hormonally up-regulated Neu-associated kinase
Gene Aliases
-
UniGene
Hs.109437
Gene Chromosome Location
Chr.21: 31873315-32004064 on Build GRCh38
Transcripts
NM_014586.1 XM_011529537.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3713406 c.470G>T p.R157L Chr.21: 31924676-31924676 on Build GRCh38
3799854 c.539C>T p.A180V Chr.21: 31924745-31924745 on Build GRCh38
1413738 c.328C>T p.R110* Chr.21: 31924534-31924534 on Build GRCh38
5680731 c.335G>A p.G112D Chr.21: 31924541-31924541 on Build GRCh38
4608940 c.526C>A p.H176N Chr.21: 31924732-31924732 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772179174 Chr.21: 31924554-31924554 on Build GRCh38
rs774902822 Chr.21: 31924685-31924685 on Build GRCh38
rs756689144 Chr.21: 31924482-31924482 on Build GRCh38
rs764591309 Chr.21: 31924776-31924776 on Build GRCh38
rs755191115 Chr.21: 31924803-31924803 on Build GRCh38
rs763925469 Chr.21: 31924465-31924465 on Build GRCh38
rs749932370 Chr.21: 31924610-31924610 on Build GRCh38
rs200760697 Chr.21: 31924777-31924777 on Build GRCh38
rs753113608 Chr.21: 31924641-31924641 on Build GRCh38
rs993341188 Chr.21: 31924420-31924420 on Build GRCh38
rs199631073 Chr.21: 31924440-31924440 on Build GRCh38
rs1010593642 Chr.21: 31924737-31924737 on Build GRCh38
rs759201238 Chr.21: 31924752-31924752 on Build GRCh38
rs545460097 Chr.21: 31924520-31924520 on Build GRCh38
rs751120837 Chr.21: 31924746-31924746 on Build GRCh38
rs769707930 Chr.21: 31924728-31924728 on Build GRCh38
rs761716649 Chr.21: 31924601-31924601 on Build GRCh38
rs756660257 Chr.21: 31924515-31924515 on Build GRCh38
rs201964145 Chr.21: 31924455-31924455 on Build GRCh38
rs13050585 Chr.21: 31924853-31924853 on Build GRCh38
rs55847614 Chr.21: 31924638-31924638 on Build GRCh38
rs752994349 Chr.21: 31924510-31924510 on Build GRCh38
rs192783090 Chr.21: 31924782-31924782 on Build GRCh38
rs538121097 Chr.21: 31924810-31924810 on Build GRCh38
rs201574800 Chr.21: 31924445-31924445 on Build GRCh38
rs745928207 Chr.21: 31924687-31924687 on Build GRCh38
rs760328005 Chr.21: 31924562-31924562 on Build GRCh38
rs779197939 Chr.21: 31924685-31924685 on Build GRCh38
rs930535095 Chr.21: 31924812-31924812 on Build GRCh38
rs138361949 Chr.21: 31924493-31924493 on Build GRCh38
rs757805328 Chr.21: 31924682-31924682 on Build GRCh38
rs906421051 Chr.21: 31924852-31924852 on Build GRCh38
rs746851498 Chr.21: 31924689-31924689 on Build GRCh38
rs781669281 Chr.21: 31924506-31924506 on Build GRCh38
rs374183829 Chr.21: 31924802-31924802 on Build GRCh38
rs369490768 Chr.21: 31924742-31924742 on Build GRCh38
rs138021652 Chr.21: 31924734-31924734 on Build GRCh38
rs768385636 Chr.21: 31924704-31924704 on Build GRCh38
rs750127108 Chr.21: 31924498-31924498 on Build GRCh38
rs564370055 Chr.21: 31924620-31924620 on Build GRCh38
rs749326655 Chr.21: 31924421-31924421 on Build GRCh38
rs374827442 Chr.21: 31924439-31924439 on Build GRCh38
rs544212060 Chr.21: 31924563-31924563 on Build GRCh38
rs760731052 Chr.21: 31924457-31924457 on Build GRCh38
rs377533874 Chr.21: 31924709-31924709 on Build GRCh38
rs745572074 Chr.21: 31924827-31924827 on Build GRCh38
rs35133981 Chr.21: 31924675-31924675 on Build GRCh38
rs758015472 Chr.21: 31924499-31924499 on Build GRCh38
rs780161504 Chr.21: 31924788-31924788 on Build GRCh38
rs779109858 Chr.21: 31924532-31924532 on Build GRCh38
rs778078613 Chr.21: 31924516-31924516 on Build GRCh38
rs746129084 Chr.21: 31924553-31924553 on Build GRCh38
rs758369859 Chr.21: 31924787-31924787 on Build GRCh38
rs768998000 Chr.21: 31924566-31924566 on Build GRCh38
rs772351266 Chr.21: 31924688-31924688 on Build GRCh38
rs189914966 Chr.21: 31924407-31924407 on Build GRCh38
rs977013844 Chr.21: 31924481-31924481 on Build GRCh38
rs532742953 Chr.21: 31924648-31924648 on Build GRCh38
rs761808304 Chr.21: 31924726-31924726 on Build GRCh38
rs962174146 Chr.21: 31924809-31924809 on Build GRCh38
rs113708542 Chr.21: 31924417-31924417 on Build GRCh38
rs150809770 Chr.21: 31924725-31924725 on Build GRCh38
rs1048135889 Chr.21: 31924846-31924846 on Build GRCh38
rs753357737 Chr.21: 31924474-31924474 on Build GRCh38
rs550429153 Chr.21: 31924819-31924819 on Build GRCh38
rs142458503 Chr.21: 31924531-31924531 on Build GRCh38
rs772725461 Chr.21: 31924740-31924740 on Build GRCh38
rs914540613 Chr.21: 31924629-31924629 on Build GRCh38
rs759652169 Chr.21: 31924412-31924412 on Build GRCh38
rs775412939 Chr.21: 31924561-31924561 on Build GRCh38
rs776504720 Chr.21: 31924590-31924590 on Build GRCh38
rs146277869 Chr.21: 31924602-31924602 on Build GRCh38
rs754441068 Chr.21: 31924676-31924676 on Build GRCh38
rs775112603 Chr.21: 31924452-31924452 on Build GRCh38
rs148302130 Chr.21: 31924632-31924632 on Build GRCh38
rs373726863 Chr.21: 31924745-31924745 on Build GRCh38

View Full Product Details