Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 52331056-52331553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH1
Entrez Gene ID
25981
Gene Name
dynein axonemal heavy chain 1
Gene Aliases
DNAHC1, HDHC7, HL-11, HL11, HSRF-1, XLHSRF-1
UniGene
Hs.655469
Gene Chromosome Location
Chr.3: 52313010-52400497 on Build GRCh38
Transcripts
NM_015512.4 XM_017006129.1 XM_017006130.1 XM_017006131.1 XM_017006132.1 XM_017006133.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6221802 c.943A>G p.K315E Chr.3: 52331219-52331219 on Build GRCh38
1485363 c.1020G>T p.G340G Chr.3: 52331296-52331296 on Build GRCh38
5416969 c.922C>G p.L308V Chr.3: 52331198-52331198 on Build GRCh38
5995125 c.909C>T p.C303C Chr.3: 52331185-52331185 on Build GRCh38
1579272 c.876C>A p.D292E Chr.3: 52331152-52331152 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765939510 Chr.3: 52331325-52331325 on Build GRCh38
rs913212438 Chr.3: 52331198-52331198 on Build GRCh38
rs778756265 Chr.3: 52331342-52331342 on Build GRCh38
rs753428890 Chr.3: 52331328-52331328 on Build GRCh38
rs764659439 Chr.3: 52331319-52331319 on Build GRCh38
rs200450328 Chr.3: 52331164-52331164 on Build GRCh38
rs73837050 Chr.3: 52331359-52331359 on Build GRCh38
rs753152874 Chr.3: 52331377-52331377 on Build GRCh38
rs557338880 Chr.3: 52331360-52331360 on Build GRCh38
rs956816383 Chr.3: 52331227-52331227 on Build GRCh38
rs773187425 Chr.3: 52331136-52331136 on Build GRCh38
rs764561677 Chr.3: 52331253-52331253 on Build GRCh38
rs565639428 Chr.3: 52331218-52331218 on Build GRCh38
rs932109765 Chr.3: 52331230-52331230 on Build GRCh38
rs1025009072 Chr.3: 52331280-52331280 on Build GRCh38
rs3864111 Chr.3: 52331245-52331245 on Build GRCh38
rs770047229 Chr.3: 52331304-52331304 on Build GRCh38
rs55931436 Chr.3: 52331207-52331207 on Build GRCh38
rs978732349 Chr.3: 52331514-52331514 on Build GRCh38
rs1039248425 Chr.3: 52331206-52331206 on Build GRCh38
rs918896087 Chr.3: 52331183-52331183 on Build GRCh38
rs765540817 Chr.3: 52331237-52331237 on Build GRCh38
rs969817976 Chr.3: 52331163-52331163 on Build GRCh38
rs773309762 Chr.3: 52331212-52331212 on Build GRCh38
rs763289186 Chr.3: 52331313-52331313 on Build GRCh38
rs867299408 Chr.3: 52331261-52331261 on Build GRCh38
rs376671317 Chr.3: 52331124-52331124 on Build GRCh38
rs186457523 Chr.3: 52331373-52331373 on Build GRCh38
rs926759884 Chr.3: 52331293-52331293 on Build GRCh38
rs758909731 Chr.3: 52331246-52331246 on Build GRCh38
rs199763713 Chr.3: 52331262-52331262 on Build GRCh38
rs766608273 Chr.3: 52331223-52331223 on Build GRCh38
rs373505739 Chr.3: 52331298-52331298 on Build GRCh38
rs935824917 Chr.3: 52331405-52331405 on Build GRCh38
rs1020147293 Chr.3: 52331443-52331443 on Build GRCh38
rs761962640 Chr.3: 52331192-52331192 on Build GRCh38
rs767128641 Chr.3: 52331133-52331135 on Build GRCh38
rs377023440 Chr.3: 52331186-52331186 on Build GRCh38
rs754175909 Chr.3: 52331168-52331168 on Build GRCh38
rs778809753 Chr.3: 52331278-52331278 on Build GRCh38
rs528031834 Chr.3: 52331205-52331205 on Build GRCh38
rs780717577 Chr.3: 52331191-52331191 on Build GRCh38
rs963552743 Chr.3: 52331295-52331295 on Build GRCh38
rs968632096 Chr.3: 52331462-52331462 on Build GRCh38
rs1035893382 Chr.3: 52331390-52331390 on Build GRCh38
rs762891487 Chr.3: 52331113-52331113 on Build GRCh38
rs749937850 Chr.3: 52331260-52331260 on Build GRCh38
rs753132213 Chr.3: 52331185-52331185 on Build GRCh38
rs747931853 Chr.3: 52331282-52331282 on Build GRCh38
rs928941028 Chr.3: 52331184-52331184 on Build GRCh38
rs559771113 Chr.3: 52331383-52331383 on Build GRCh38
rs201423629 Chr.3: 52331112-52331112 on Build GRCh38
rs368468783 Chr.3: 52331284-52331284 on Build GRCh38
rs749092447 Chr.3: 52331199-52331199 on Build GRCh38
rs988324191 Chr.3: 52331112-52331113 on Build GRCh38
rs371897357 Chr.3: 52331324-52331324 on Build GRCh38
rs777073051 Chr.3: 52331115-52331115 on Build GRCh38
rs751945958 Chr.3: 52331323-52331323 on Build GRCh38
rs755640904 Chr.3: 52331210-52331212 on Build GRCh38
rs768499149 Chr.3: 52331201-52331201 on Build GRCh38
rs201633397 Chr.3: 52331290-52331290 on Build GRCh38
rs754698653 Chr.3: 52331275-52331275 on Build GRCh38
rs775812913 Chr.3: 52331306-52331306 on Build GRCh38
rs748892873 Chr.3: 52331273-52331273 on Build GRCh38
rs193242312 Chr.3: 52331172-52331172 on Build GRCh38
rs145999578 Chr.3: 52331433-52331433 on Build GRCh38
rs759717790 Chr.3: 52331303-52331303 on Build GRCh38
rs1052358371 Chr.3: 52331236-52331236 on Build GRCh38
rs776701745 Chr.3: 52331300-52331300 on Build GRCh38
rs779912625 Chr.3: 52331195-52331195 on Build GRCh38
rs987535202 Chr.3: 52331331-52331331 on Build GRCh38
rs1038971632 Chr.3: 52331382-52331382 on Build GRCh38
rs369216877 Chr.3: 52331194-52331194 on Build GRCh38
rs776900676 Chr.3: 52331228-52331228 on Build GRCh38
rs771932160 Chr.3: 52331210-52331210 on Build GRCh38
rs577271293 Chr.3: 52331369-52331369 on Build GRCh38
rs199505182 Chr.3: 52331209-52331209 on Build GRCh38
rs989107148 Chr.3: 52331435-52331435 on Build GRCh38
rs769364888 Chr.3: 52331100-52331100 on Build GRCh38
rs188504147 Chr.3: 52331162-52331162 on Build GRCh38
rs185728699 Chr.3: 52331329-52331329 on Build GRCh38
rs143657729 Chr.3: 52331133-52331133 on Build GRCh38
rs752416979 Chr.3: 52331177-52331177 on Build GRCh38
rs755391323 Chr.3: 52331170-52331170 on Build GRCh38
rs370393988 Chr.3: 52331229-52331229 on Build GRCh38

View Full Product Details