Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 127805552-127806072 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA4L
Entrez Gene ID
22824
Gene Name
heat shock protein family A (Hsp70) member 4 like
Gene Aliases
APG-1, HSPH3, Osp94
UniGene
Hs.135554
Gene Chromosome Location
Chr.4: 127781821-127834073 on Build GRCh38
Transcripts
NM_001317381.1 NM_001317382.1 NM_001317383.1 NM_014278.3 XM_011531745.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4122175 c.1235C>T p.A412V Chr.4: 127805691-127805691 on Build GRCh38
3825168 c.1236G>A p.A412A Chr.4: 127805692-127805692 on Build GRCh38
4893229 c.1331G>A p.G444E Chr.4: 127805787-127805787 on Build GRCh38
1051100 c.1156G>A p.A386T Chr.4: 127805705-127805705 on Build GRCh38
3825167 c.1143G>A p.A381A Chr.4: 127805692-127805692 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896271076 Chr.4: 127805958-127805958 on Build GRCh38
rs746183485 Chr.4: 127805814-127805814 on Build GRCh38
rs762956801 Chr.4: 127805842-127805842 on Build GRCh38
rs906074723 Chr.4: 127805628-127805628 on Build GRCh38
rs941368045 Chr.4: 127805920-127805921 on Build GRCh38
rs1038734993 Chr.4: 127805960-127805960 on Build GRCh38
rs577260042 Chr.4: 127805692-127805692 on Build GRCh38
rs373956376 Chr.4: 127805843-127805843 on Build GRCh38
rs150334200 Chr.4: 127805680-127805680 on Build GRCh38
rs935013384 Chr.4: 127805954-127805954 on Build GRCh38
rs751820980 Chr.4: 127805640-127805640 on Build GRCh38
rs377432911 Chr.4: 127805721-127805721 on Build GRCh38
rs768329117 Chr.4: 127805820-127805820 on Build GRCh38
rs542465014 Chr.4: 127805989-127805989 on Build GRCh38
rs979392343 Chr.4: 127805807-127805807 on Build GRCh38
rs771226189 Chr.4: 127805685-127805685 on Build GRCh38
rs149774219 Chr.4: 127805745-127805745 on Build GRCh38
rs575054744 Chr.4: 127805985-127805985 on Build GRCh38
rs770507380 Chr.4: 127805937-127805937 on Build GRCh38
rs780440502 Chr.4: 127805638-127805638 on Build GRCh38
rs376474359 Chr.4: 127805821-127805821 on Build GRCh38
rs749140580 Chr.4: 127805625-127805626 on Build GRCh38
rs768433602 Chr.4: 127805817-127805817 on Build GRCh38
rs751418054 Chr.4: 127805845-127805845 on Build GRCh38
rs1052180768 Chr.4: 127805957-127805957 on Build GRCh38
rs533891706 Chr.4: 127805668-127805668 on Build GRCh38
rs1027587444 Chr.4: 127805642-127805642 on Build GRCh38
rs769176639 Chr.4: 127805838-127805838 on Build GRCh38
rs1024551783 Chr.4: 127805607-127805607 on Build GRCh38
rs202039142 Chr.4: 127805666-127805666 on Build GRCh38
rs761402132 Chr.4: 127805823-127805823 on Build GRCh38
rs906047668 Chr.4: 127805996-127805996 on Build GRCh38
rs931852692 Chr.4: 127805793-127805793 on Build GRCh38
rs779217447 Chr.4: 127805811-127805811 on Build GRCh38
rs1002163288 Chr.4: 127806023-127806023 on Build GRCh38
rs368907722 Chr.4: 127805744-127805744 on Build GRCh38
rs766274911 Chr.4: 127805729-127805729 on Build GRCh38
rs771351099 Chr.4: 127805804-127805804 on Build GRCh38
rs775179109 Chr.4: 127805687-127805687 on Build GRCh38
rs1011695390 Chr.4: 127805586-127805586 on Build GRCh38
rs773270794 Chr.4: 127805839-127805839 on Build GRCh38
rs111634215 Chr.4: 127805935-127805935 on Build GRCh38
rs578208205 Chr.4: 127805913-127805913 on Build GRCh38
rs749781458 Chr.4: 127805769-127805769 on Build GRCh38
rs1027888231 Chr.4: 127806036-127806036 on Build GRCh38
rs761190089 Chr.4: 127805713-127805713 on Build GRCh38
rs556463415 Chr.4: 127805975-127805975 on Build GRCh38
rs976356893 Chr.4: 127805747-127805747 on Build GRCh38
rs934846128 Chr.4: 127805991-127805991 on Build GRCh38
rs751873781 Chr.4: 127805733-127805733 on Build GRCh38
rs776211650 Chr.4: 127805835-127805850 on Build GRCh38
rs139673474 Chr.4: 127805717-127805717 on Build GRCh38
rs386679390 Chr.4: 127805897-127805898 on Build GRCh38
rs748924245 Chr.4: 127805816-127805816 on Build GRCh38
rs374327031 Chr.4: 127805718-127805718 on Build GRCh38
rs755179832 Chr.4: 127805665-127805665 on Build GRCh38
rs1046029952 Chr.4: 127805986-127805986 on Build GRCh38
rs1052850003 Chr.4: 127806000-127806000 on Build GRCh38
rs907353138 Chr.4: 127805987-127805987 on Build GRCh38
rs781218367 Chr.4: 127805743-127805743 on Build GRCh38
rs761436359 Chr.4: 127805840-127805840 on Build GRCh38
rs546695454 Chr.4: 127805877-127805877 on Build GRCh38
rs768194196 Chr.4: 127805691-127805691 on Build GRCh38
rs915919607 Chr.4: 127805980-127805980 on Build GRCh38
rs755232976 Chr.4: 127805739-127805739 on Build GRCh38
rs778099201 Chr.4: 127805673-127805673 on Build GRCh38
rs778355069 Chr.4: 127805753-127805753 on Build GRCh38
rs924911988 Chr.4: 127805824-127805824 on Build GRCh38

View Full Product Details