Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150810371-150810904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARNT
Entrez Gene ID
405
Gene Name
aryl hydrocarbon receptor nuclear translocator
Gene Aliases
HIF-1-beta, HIF-1beta, HIF1-beta, HIF1B, HIF1BETA, TANGO, bHLHe2
UniGene
Hs.632446
Gene Chromosome Location
Chr.1: 150809705-150876768 on Build GRCh38
Transcripts
NM_001197325.1 NM_001286035.1 NM_001286036.1 NM_001668.3 NM_178427.2 XM_005245151.1 XM_005245153.1 XM_005245154.3 XM_005245157.1 XM_011509542.2 XM_011509543.2 XM_011509544.2 XM_011509545.2 XM_011509546.1 XM_011509547.2 XM_017001288.1 XM_017001289.1 XM_017001290.1 XM_017001291.1 XM_017001292.1 XM_017001293.1 XM_017001294.1 XM_017001295.1 XM_017001296.1

SNP Detail

SNP ID Chromosome Location
rs748669896 Chr.1: 150810826-150810826 on Build GRCh38
rs763088080 Chr.1: 150810702-150810702 on Build GRCh38
rs61817634 Chr.1: 150810630-150810630 on Build GRCh38
rs71645814 Chr.1: 150810633-150810633 on Build GRCh38
rs181800295 Chr.1: 150810767-150810767 on Build GRCh38
rs1000930277 Chr.1: 150810412-150810412 on Build GRCh38
rs946255950 Chr.1: 150810812-150810812 on Build GRCh38
rs780354320 Chr.1: 150810419-150810419 on Build GRCh38
rs922211917 Chr.1: 150810483-150810483 on Build GRCh38
rs869246025 Chr.1: 150810648-150810650 on Build GRCh38
rs796477601 Chr.1: 150810695-150810695 on Build GRCh38
rs929126414 Chr.1: 150810677-150810677 on Build GRCh38
rs904532090 Chr.1: 150810765-150810765 on Build GRCh38
rs970273219 Chr.1: 150810852-150810852 on Build GRCh38
rs1032223253 Chr.1: 150810504-150810504 on Build GRCh38
rs1038105699 Chr.1: 150810465-150810465 on Build GRCh38
rs35400828 Chr.1: 150810503-150810504 on Build GRCh38
rs912133981 Chr.1: 150810781-150810781 on Build GRCh38
rs1017985303 Chr.1: 150810747-150810747 on Build GRCh38
rs1032387947 Chr.1: 150810632-150810633 on Build GRCh38
rs587612751 Chr.1: 150810646-150810646 on Build GRCh38
rs966374487 Chr.1: 150810400-150810400 on Build GRCh38
rs369972465 Chr.1: 150810576-150810580 on Build GRCh38
rs917487892 Chr.1: 150810555-150810555 on Build GRCh38
rs10305752 Chr.1: 150810763-150810764 on Build GRCh38
rs796584528 Chr.1: 150810762-150810763 on Build GRCh38
rs755379548 Chr.1: 150810762-150810762 on Build GRCh38
rs796837180 Chr.1: 150810721-150810722 on Build GRCh38
rs902277365 Chr.1: 150810825-150810825 on Build GRCh38
rs990673449 Chr.1: 150810420-150810423 on Build GRCh38
rs374493943 Chr.1: 150810662-150810662 on Build GRCh38
rs963418658 Chr.1: 150810704-150810704 on Build GRCh38
rs587738877 Chr.1: 150810632-150810632 on Build GRCh38
rs1051953782 Chr.1: 150810539-150810539 on Build GRCh38
rs746446716 Chr.1: 150810701-150810701 on Build GRCh38
rs973436786 Chr.1: 150810710-150810710 on Build GRCh38
rs115569224 Chr.1: 150810690-150810690 on Build GRCh38
rs932272896 Chr.1: 150810744-150810744 on Build GRCh38
rs956185280 Chr.1: 150810534-150810534 on Build GRCh38
rs1053473808 Chr.1: 150810832-150810832 on Build GRCh38
rs894826989 Chr.1: 150810836-150810836 on Build GRCh38
rs970819675 Chr.1: 150810438-150810439 on Build GRCh38
rs926325873 Chr.1: 150810628-150810628 on Build GRCh38
rs942044860 Chr.1: 150810451-150810451 on Build GRCh38
rs147463981 Chr.1: 150810816-150810816 on Build GRCh38
rs993035365 Chr.1: 150810550-150810550 on Build GRCh38
rs978872522 Chr.1: 150810440-150810440 on Build GRCh38
rs190599470 Chr.1: 150810709-150810709 on Build GRCh38
rs10305754 Chr.1: 150810663-150810663 on Build GRCh38
rs948978414 Chr.1: 150810562-150810562 on Build GRCh38
rs10305753 Chr.1: 150810722-150810723 on Build GRCh38
rs1010343836 Chr.1: 150810565-150810565 on Build GRCh38
rs897611427 Chr.1: 150810674-150810674 on Build GRCh38
rs10305755 Chr.1: 150810566-150810566 on Build GRCh38
rs917714533 Chr.1: 150810435-150810435 on Build GRCh38
rs879057331 Chr.1: 150810633-150810634 on Build GRCh38
rs1000538210 Chr.1: 150810587-150810587 on Build GRCh38
rs587627934 Chr.1: 150810857-150810857 on Build GRCh38
rs907945570 Chr.1: 150810446-150810446 on Build GRCh38
rs1041827290 Chr.1: 150810815-150810815 on Build GRCh38
rs1031753349 Chr.1: 150810780-150810780 on Build GRCh38
rs941733714 Chr.1: 150810650-150810650 on Build GRCh38
rs959329845 Chr.1: 150810633-150810633 on Build GRCh38
rs956131404 Chr.1: 150810792-150810792 on Build GRCh38
rs1032297835 Chr.1: 150810418-150810418 on Build GRCh38
rs1051577207 Chr.1: 150810678-150810678 on Build GRCh38
rs1007450611 Chr.1: 150810692-150810692 on Build GRCh38

View Full Product Details