Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 18661855-18662360 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTSL1
Entrez Gene ID
92949
Gene Name
ADAMTS like 1
Gene Aliases
ADAMTSL-1, ADAMTSR1, C9orf94, PUNCTIN
UniGene
Hs.741483
Gene Chromosome Location
Chr.9: 17906574-18910950 on Build GRCh38
Transcripts
NM_001040272.5 NM_052866.4 XM_011518063.2 XM_011518064.2 XM_011518067.1 XM_017015310.1 XM_017015311.1 XM_017015312.1 XM_017015313.1 XM_017015314.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
158913 c.1012T>A p.C338S Chr.9: 18662000-18662000 on Build GRCh38
3745942 c.1069G>A p.D357N Chr.9: 18662057-18662057 on Build GRCh38
6991485 c.1002T>C p.A334A Chr.9: 18661990-18661990 on Build GRCh38
5101483 c.997G>A p.V333I Chr.9: 18661985-18661985 on Build GRCh38
3731041 c.1001C>T p.A334V Chr.9: 18661989-18661989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1001127356 Chr.9: 18662260-18662260 on Build GRCh38
rs571356973 Chr.9: 18662322-18662322 on Build GRCh38
rs563794441 Chr.9: 18662114-18662114 on Build GRCh38
rs987282681 Chr.9: 18661988-18661988 on Build GRCh38
rs1034844192 Chr.9: 18662149-18662149 on Build GRCh38
rs377552651 Chr.9: 18662081-18662081 on Build GRCh38
rs749267542 Chr.9: 18662015-18662015 on Build GRCh38
rs1003406001 Chr.9: 18662146-18662146 on Build GRCh38
rs184527839 Chr.9: 18662150-18662150 on Build GRCh38
rs770603282 Chr.9: 18661942-18661942 on Build GRCh38
rs118100207 Chr.9: 18662268-18662268 on Build GRCh38
rs370253600 Chr.9: 18661974-18661974 on Build GRCh38
rs763773238 Chr.9: 18661888-18661888 on Build GRCh38
rs777583332 Chr.9: 18661950-18661950 on Build GRCh38
rs145178068 Chr.9: 18662284-18662284 on Build GRCh38
rs893485736 Chr.9: 18662266-18662266 on Build GRCh38
rs141627942 Chr.9: 18661963-18661963 on Build GRCh38
rs73433591 Chr.9: 18662014-18662014 on Build GRCh38
rs911386715 Chr.9: 18662235-18662235 on Build GRCh38
rs777106613 Chr.9: 18661981-18661981 on Build GRCh38
rs1017208369 Chr.9: 18661886-18661886 on Build GRCh38
rs748749844 Chr.9: 18661967-18661967 on Build GRCh38
rs148178041 Chr.9: 18661951-18661951 on Build GRCh38
rs918352651 Chr.9: 18662147-18662147 on Build GRCh38
rs934136209 Chr.9: 18661998-18661998 on Build GRCh38
rs763501571 Chr.9: 18662088-18662088 on Build GRCh38
rs1056205703 Chr.9: 18662152-18662152 on Build GRCh38
rs751760171 Chr.9: 18661990-18661990 on Build GRCh38
rs776771 Chr.9: 18662294-18662294 on Build GRCh38
rs745610882 Chr.9: 18662013-18662013 on Build GRCh38
rs752837790 Chr.9: 18661918-18661918 on Build GRCh38
rs896869483 Chr.9: 18661894-18661894 on Build GRCh38
rs142112150 Chr.9: 18661980-18661980 on Build GRCh38
rs776143369 Chr.9: 18662204-18662204 on Build GRCh38
rs985438002 Chr.9: 18662334-18662334 on Build GRCh38
rs780660664 Chr.9: 18662169-18662169 on Build GRCh38
rs1039159495 Chr.9: 18662257-18662257 on Build GRCh38
rs748846167 Chr.9: 18662102-18662102 on Build GRCh38
rs150088929 Chr.9: 18661979-18661979 on Build GRCh38
rs747739409 Chr.9: 18662030-18662030 on Build GRCh38
rs986156040 Chr.9: 18662034-18662034 on Build GRCh38
rs1016946086 Chr.9: 18662185-18662185 on Build GRCh38
rs973362151 Chr.9: 18661993-18661993 on Build GRCh38
rs376273773 Chr.9: 18661948-18661948 on Build GRCh38
rs546597555 Chr.9: 18662218-18662218 on Build GRCh38
rs1011906000 Chr.9: 18662210-18662210 on Build GRCh38
rs764147499 Chr.9: 18662107-18662107 on Build GRCh38
rs902412890 Chr.9: 18662144-18662144 on Build GRCh38
rs537641249 Chr.9: 18661934-18661934 on Build GRCh38
rs574508977 Chr.9: 18661982-18661982 on Build GRCh38
rs558928988 Chr.9: 18661941-18661941 on Build GRCh38
rs963007007 Chr.9: 18662186-18662186 on Build GRCh38
rs943123910 Chr.9: 18662241-18662241 on Build GRCh38
rs191918530 Chr.9: 18662036-18662036 on Build GRCh38
rs746762586 Chr.9: 18662085-18662085 on Build GRCh38
rs765624574 Chr.9: 18662067-18662067 on Build GRCh38
rs759899921 Chr.9: 18662062-18662062 on Build GRCh38
rs758162162 Chr.9: 18661929-18661929 on Build GRCh38
rs1003460145 Chr.9: 18662289-18662289 on Build GRCh38
rs141659739 Chr.9: 18661997-18661997 on Build GRCh38
rs771820042 Chr.9: 18662035-18662035 on Build GRCh38
rs373835128 Chr.9: 18661913-18661913 on Build GRCh38
rs764202410 Chr.9: 18661985-18661985 on Build GRCh38
rs184592772 Chr.9: 18661895-18661895 on Build GRCh38
rs200908366 Chr.9: 18661896-18661896 on Build GRCh38
rs757559183 Chr.9: 18661994-18661994 on Build GRCh38
rs140226148 Chr.9: 18662254-18662254 on Build GRCh38
rs139287759 Chr.9: 18662020-18662020 on Build GRCh38
rs941527676 Chr.9: 18662127-18662127 on Build GRCh38
rs528363799 Chr.9: 18662188-18662188 on Build GRCh38
rs370486688 Chr.9: 18662115-18662115 on Build GRCh38
rs568967066 Chr.9: 18662276-18662276 on Build GRCh38
rs756246381 Chr.9: 18661964-18661964 on Build GRCh38
rs753230182 Chr.9: 18661984-18661984 on Build GRCh38
rs896191168 Chr.9: 18662184-18662184 on Build GRCh38
rs1038736765 Chr.9: 18662052-18662052 on Build GRCh38
rs775947988 Chr.9: 18662068-18662068 on Build GRCh38
rs903731462 Chr.9: 18662227-18662227 on Build GRCh38
rs775310518 Chr.9: 18662047-18662047 on Build GRCh38
rs564157493 Chr.9: 18662155-18662155 on Build GRCh38
rs754266647 Chr.9: 18661909-18661909 on Build GRCh38
rs755224993 Chr.9: 18661890-18661890 on Build GRCh38
rs548252326 Chr.9: 18662143-18662143 on Build GRCh38
rs990865204 Chr.9: 18662182-18662182 on Build GRCh38
rs973029193 Chr.9: 18662199-18662199 on Build GRCh38
rs761371380 Chr.9: 18662219-18662219 on Build GRCh38
rs757289369 Chr.9: 18662123-18662123 on Build GRCh38
rs768249524 Chr.9: 18662018-18662018 on Build GRCh38
rs774106541 Chr.9: 18661975-18661975 on Build GRCh38
rs758490070 Chr.9: 18661928-18661928 on Build GRCh38
rs550644423 Chr.9: 18662269-18662269 on Build GRCh38

View Full Product Details