Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 136308689-136309189 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRG4
Entrez Gene ID
139378
Gene Name
adhesion G protein-coupled receptor G4
Gene Aliases
GPR112, PGR17, RP1-299I16
UniGene
Hs.381354
Gene Chromosome Location
Chr.X: 136300963-136416888 on Build GRCh38
Transcripts
NM_153834.3 XM_011531269.2 XM_011531271.2

SNP Detail

SNP ID Chromosome Location
rs766781235 Chr.X: 136308879-136308879 on Build GRCh38
rs745822568 Chr.X: 136308745-136308745 on Build GRCh38
rs977186054 Chr.X: 136309048-136309048 on Build GRCh38
rs749939590 Chr.X: 136308838-136308838 on Build GRCh38
rs762779561 Chr.X: 136308822-136308822 on Build GRCh38
rs746133878 Chr.X: 136308741-136308741 on Build GRCh38
rs755390655 Chr.X: 136308881-136308881 on Build GRCh38
rs367694564 Chr.X: 136309019-136309019 on Build GRCh38
rs766874331 Chr.X: 136308870-136308870 on Build GRCh38
rs755511589 Chr.X: 136308931-136308931 on Build GRCh38
rs764285537 Chr.X: 136308840-136308843 on Build GRCh38
rs750727669 Chr.X: 136308823-136308823 on Build GRCh38
rs894826241 Chr.X: 136308788-136308788 on Build GRCh38
rs1003345681 Chr.X: 136308789-136308791 on Build GRCh38
rs1000492320 Chr.X: 136308989-136308989 on Build GRCh38
rs756719617 Chr.X: 136309065-136309065 on Build GRCh38
rs767149317 Chr.X: 136308836-136308836 on Build GRCh38
rs773213079 Chr.X: 136308743-136308743 on Build GRCh38
rs769706208 Chr.X: 136308795-136308795 on Build GRCh38
rs774082737 Chr.X: 136308825-136308825 on Build GRCh38
rs374858229 Chr.X: 136308827-136308827 on Build GRCh38
rs772172872 Chr.X: 136308727-136308727 on Build GRCh38
rs983242545 Chr.X: 136309131-136309131 on Build GRCh38
rs373230154 Chr.X: 136308871-136308871 on Build GRCh38
rs748761791 Chr.X: 136309057-136309057 on Build GRCh38
rs1017667270 Chr.X: 136309012-136309012 on Build GRCh38
rs951480162 Chr.X: 136309123-136309123 on Build GRCh38
rs765387392 Chr.X: 136308820-136308820 on Build GRCh38
rs369417179 Chr.X: 136308828-136308828 on Build GRCh38
rs1031170293 Chr.X: 136308924-136308924 on Build GRCh38
rs964059332 Chr.X: 136309040-136309040 on Build GRCh38
rs762340721 Chr.X: 136309056-136309056 on Build GRCh38
rs749685149 Chr.X: 136308857-136308858 on Build GRCh38
rs377314366 Chr.X: 136308901-136308901 on Build GRCh38
rs747373980 Chr.X: 136308716-136308716 on Build GRCh38
rs999642479 Chr.X: 136308913-136308913 on Build GRCh38
rs1011121047 Chr.X: 136308946-136308946 on Build GRCh38
rs775909714 Chr.X: 136308714-136308714 on Build GRCh38
rs769261208 Chr.X: 136308860-136308861 on Build GRCh38
rs976226177 Chr.X: 136309060-136309060 on Build GRCh38
rs756747355 Chr.X: 136308859-136308859 on Build GRCh38
rs780240741 Chr.X: 136308852-136308853 on Build GRCh38
rs762245498 Chr.X: 136308778-136308778 on Build GRCh38

View Full Product Details