Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74339157-74339650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP11A1
Entrez Gene ID
1583
Gene Name
cytochrome P450 family 11 subfamily A member 1
Gene Aliases
CYP11A, CYPXIA1, P450SCC
UniGene
Hs.303980
Gene Chromosome Location
Chr.15: 74337762-74367740 on Build GRCh38
Transcripts
NM_000781.2 NM_001099773.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1938457 c.1102G>A p.A368T Chr.15: 74339642-74339642 on Build GRCh38
6638992 c.1099A>G p.M367V Chr.15: 74339645-74339645 on Build GRCh38
290569 c.1173C>T p.S391S Chr.15: 74339300-74339300 on Build GRCh38
5888251 c.1129C>T p.L377F Chr.15: 74339615-74339615 on Build GRCh38
6638990 c.1236+2T>C p.? Chr.15: 74339235-74339235 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771539144 Chr.15: 74339334-74339334 on Build GRCh38
rs756852433 Chr.15: 74339557-74339558 on Build GRCh38
rs1031216597 Chr.15: 74339529-74339529 on Build GRCh38
rs761496946 Chr.15: 74339222-74339222 on Build GRCh38
rs1001293993 Chr.15: 74339576-74339576 on Build GRCh38
rs146628086 Chr.15: 74339338-74339338 on Build GRCh38
rs756410813 Chr.15: 74339567-74339567 on Build GRCh38
rs759453073 Chr.15: 74339277-74339277 on Build GRCh38
rs751841383 Chr.15: 74339542-74339542 on Build GRCh38
rs886051479 Chr.15: 74339272-74339272 on Build GRCh38
rs1049909813 Chr.15: 74339186-74339186 on Build GRCh38
rs1012890303 Chr.15: 74339516-74339516 on Build GRCh38
rs771663597 Chr.15: 74339247-74339247 on Build GRCh38
rs760502211 Chr.15: 74339252-74339252 on Build GRCh38
rs1025373899 Chr.15: 74339417-74339418 on Build GRCh38
rs537187397 Chr.15: 74339309-74339309 on Build GRCh38
rs112737388 Chr.15: 74339340-74339340 on Build GRCh38
rs369183062 Chr.15: 74339328-74339328 on Build GRCh38
rs768022753 Chr.15: 74339562-74339562 on Build GRCh38
rs577204139 Chr.15: 74339524-74339524 on Build GRCh38
rs746361914 Chr.15: 74339318-74339318 on Build GRCh38
rs1041074789 Chr.15: 74339269-74339269 on Build GRCh38
rs757483896 Chr.15: 74339558-74339558 on Build GRCh38
rs746607026 Chr.15: 74339342-74339342 on Build GRCh38
rs555201959 Chr.15: 74339419-74339419 on Build GRCh38
rs894165406 Chr.15: 74339513-74339513 on Build GRCh38
rs944254592 Chr.15: 74339485-74339485 on Build GRCh38
rs1043137431 Chr.15: 74339563-74339563 on Build GRCh38
rs766208059 Chr.15: 74339400-74339400 on Build GRCh38
rs565795598 Chr.15: 74339368-74339368 on Build GRCh38
rs763197214 Chr.15: 74339367-74339367 on Build GRCh38
rs776126810 Chr.15: 74339625-74339625 on Build GRCh38
rs895385110 Chr.15: 74339184-74339184 on Build GRCh38
rs773840614 Chr.15: 74339213-74339213 on Build GRCh38
rs764452686 Chr.15: 74339299-74339299 on Build GRCh38
rs920346701 Chr.15: 74339436-74339436 on Build GRCh38
rs149219677 Chr.15: 74339371-74339371 on Build GRCh38
rs1034148229 Chr.15: 74339582-74339582 on Build GRCh38
rs751072562 Chr.15: 74339523-74339523 on Build GRCh38
rs754662184 Chr.15: 74339526-74339526 on Build GRCh38
rs770717461 Chr.15: 74339351-74339351 on Build GRCh38
rs1036035125 Chr.15: 74339429-74339429 on Build GRCh38
rs762596641 Chr.15: 74339522-74339522 on Build GRCh38
rs750350590 Chr.15: 74339385-74339385 on Build GRCh38
rs867506250 Chr.15: 74339311-74339311 on Build GRCh38
rs895672441 Chr.15: 74339622-74339622 on Build GRCh38
rs746369410 Chr.15: 74339211-74339211 on Build GRCh38
rs201056892 Chr.15: 74339329-74339329 on Build GRCh38
rs879479808 Chr.15: 74339478-74339478 on Build GRCh38
rs375743628 Chr.15: 74339291-74339291 on Build GRCh38
rs1055860457 Chr.15: 74339509-74339509 on Build GRCh38
rs761400617 Chr.15: 74339229-74339229 on Build GRCh38
rs772829617 Chr.15: 74339249-74339249 on Build GRCh38
rs751829411 Chr.15: 74339300-74339300 on Build GRCh38
rs992748565 Chr.15: 74339401-74339401 on Build GRCh38
rs143006315 Chr.15: 74339564-74339564 on Build GRCh38
rs757960246 Chr.15: 74339591-74339591 on Build GRCh38
rs200029503 Chr.15: 74339278-74339278 on Build GRCh38
rs759416343 Chr.15: 74339360-74339360 on Build GRCh38
rs890033896 Chr.15: 74339193-74339193 on Build GRCh38
rs138177167 Chr.15: 74339306-74339306 on Build GRCh38
rs143743827 Chr.15: 74339598-74339598 on Build GRCh38
rs780648305 Chr.15: 74339571-74339571 on Build GRCh38
rs749754082 Chr.15: 74339585-74339585 on Build GRCh38
rs548790439 Chr.15: 74339199-74339199 on Build GRCh38
rs776448335 Chr.15: 74339359-74339359 on Build GRCh38
rs537382499 Chr.15: 74339443-74339443 on Build GRCh38
rs536323741 Chr.15: 74339418-74339418 on Build GRCh38
rs753788398 Chr.15: 74339270-74339270 on Build GRCh38
rs746694870 Chr.15: 74339609-74339609 on Build GRCh38
rs911701028 Chr.15: 74339421-74339421 on Build GRCh38
rs945717675 Chr.15: 74339555-74339555 on Build GRCh38
rs559297406 Chr.15: 74339620-74339620 on Build GRCh38
rs766156714 Chr.15: 74339267-74339267 on Build GRCh38
rs764260204 Chr.15: 74339539-74339539 on Build GRCh38
rs768062508 Chr.15: 74339212-74339212 on Build GRCh38
rs940539396 Chr.15: 74339426-74339426 on Build GRCh38
rs931435859 Chr.15: 74339425-74339425 on Build GRCh38
rs576536357 Chr.15: 74339437-74339437 on Build GRCh38
rs944327325 Chr.15: 74339257-74339257 on Build GRCh38
rs372603375 Chr.15: 74339319-74339319 on Build GRCh38
rs1001368906 Chr.15: 74339370-74339370 on Build GRCh38
rs377242922 Chr.15: 74339320-74339320 on Build GRCh38
rs201930933 Chr.15: 74339611-74339611 on Build GRCh38
rs559198797 Chr.15: 74339504-74339504 on Build GRCh38
rs775262462 Chr.15: 74339363-74339363 on Build GRCh38
rs752776256 Chr.15: 74339290-74339290 on Build GRCh38

View Full Product Details