Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 172951426-172951921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERGIC1
Entrez Gene ID
57222
Gene Name
endoplasmic reticulum-golgi intermediate compartment 1
Gene Aliases
ERGIC-32, ERGIC32, NET24
UniGene
Hs.509163
Gene Chromosome Location
Chr.5: 172834220-172952685 on Build GRCh38
Transcripts
NM_001031711.2 XM_006714892.1 XM_011534597.1

SNP Detail

SNP ID Chromosome Location
rs763184236 Chr.5: 172951884-172951884 on Build GRCh38
rs940715669 Chr.5: 172951721-172951721 on Build GRCh38
rs572848468 Chr.5: 172951765-172951765 on Build GRCh38
rs949545762 Chr.5: 172951493-172951493 on Build GRCh38
rs764400185 Chr.5: 172951889-172951889 on Build GRCh38
rs386695023 Chr.5: 172951853-172951854 on Build GRCh38
rs149464075 Chr.5: 172951866-172951866 on Build GRCh38
rs184199976 Chr.5: 172951488-172951488 on Build GRCh38
rs988277927 Chr.5: 172951481-172951481 on Build GRCh38
rs928012740 Chr.5: 172951706-172951706 on Build GRCh38
rs770875871 Chr.5: 172951712-172951712 on Build GRCh38
rs745939111 Chr.5: 172951805-172951805 on Build GRCh38
rs113014301 Chr.5: 172951574-172951574 on Build GRCh38
rs1009805628 Chr.5: 172951798-172951798 on Build GRCh38
rs886975707 Chr.5: 172951679-172951679 on Build GRCh38
rs577205775 Chr.5: 172951627-172951627 on Build GRCh38
rs539936 Chr.5: 172951854-172951854 on Build GRCh38
rs989752430 Chr.5: 172951804-172951804 on Build GRCh38
rs897107176 Chr.5: 172951737-172951737 on Build GRCh38
rs1008465409 Chr.5: 172951699-172951699 on Build GRCh38
rs1036356793 Chr.5: 172951863-172951863 on Build GRCh38
rs931177898 Chr.5: 172951667-172951667 on Build GRCh38
rs778917889 Chr.5: 172951737-172951737 on Build GRCh38
rs1055839510 Chr.5: 172951631-172951631 on Build GRCh38
rs1019378781 Chr.5: 172951743-172951743 on Build GRCh38
rs532677842 Chr.5: 172951857-172951857 on Build GRCh38
rs574963784 Chr.5: 172951795-172951795 on Build GRCh38
rs914109312 Chr.5: 172951492-172951492 on Build GRCh38
rs184675073 Chr.5: 172951786-172951786 on Build GRCh38
rs958256526 Chr.5: 172951465-172951465 on Build GRCh38
rs746956968 Chr.5: 172951608-172951608 on Build GRCh38
rs1046485341 Chr.5: 172951678-172951678 on Build GRCh38
rs1007107615 Chr.5: 172951739-172951739 on Build GRCh38
rs996872865 Chr.5: 172951767-172951767 on Build GRCh38
rs529359184 Chr.5: 172951724-172951724 on Build GRCh38
rs970941290 Chr.5: 172951822-172951822 on Build GRCh38
rs866775320 Chr.5: 172951532-172951532 on Build GRCh38
rs958313646 Chr.5: 172951802-172951802 on Build GRCh38
rs536403401 Chr.5: 172951738-172951738 on Build GRCh38
rs11556501 Chr.5: 172951773-172951773 on Build GRCh38
rs60165841 Chr.5: 172951853-172951853 on Build GRCh38
rs137939949 Chr.5: 172951785-172951785 on Build GRCh38
rs562255010 Chr.5: 172951560-172951560 on Build GRCh38
rs192765437 Chr.5: 172951643-172951643 on Build GRCh38
rs769797759 Chr.5: 172951851-172951851 on Build GRCh38
rs868598986 Chr.5: 172951885-172951885 on Build GRCh38
rs372493396 Chr.5: 172951817-172951817 on Build GRCh38
rs187579947 Chr.5: 172951547-172951547 on Build GRCh38
rs982664470 Chr.5: 172951838-172951838 on Build GRCh38
rs777577658 Chr.5: 172951607-172951607 on Build GRCh38
rs537233269 Chr.5: 172951540-172951540 on Build GRCh38
rs1047011416 Chr.5: 172951670-172951670 on Build GRCh38

View Full Product Details