Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 11919558-11920072 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD3
Entrez Gene ID
22903
Gene Name
BTB domain containing 3
Gene Aliases
dJ742J24.1
UniGene
Hs.709366
Gene Chromosome Location
Chr.20: 11890723-11926595 on Build GRCh38
Transcripts
NM_001282550.1 NM_001282551.1 NM_001282552.2 NM_001282554.1 NM_014962.3 NM_181443.3 XM_017027728.1 XM_017027729.1 XM_017027730.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
443454 c.452C>T p.A151V Chr.20: 11919752-11919752 on Build GRCh38
5731943 c.475G>A p.E159K Chr.20: 11919775-11919775 on Build GRCh38
6613364 c.506T>C p.V169A Chr.20: 11919806-11919806 on Build GRCh38
1410360 c.471T>A p.L157L Chr.20: 11919771-11919771 on Build GRCh38
317057 c.452C>G p.A151G Chr.20: 11919752-11919752 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748478643 Chr.20: 11919842-11919842 on Build GRCh38
rs1036868221 Chr.20: 11919706-11919706 on Build GRCh38
rs1010828066 Chr.20: 11920020-11920020 on Build GRCh38
rs368759423 Chr.20: 11919807-11919807 on Build GRCh38
rs746183579 Chr.20: 11919614-11919614 on Build GRCh38
rs984836799 Chr.20: 11919957-11919957 on Build GRCh38
rs539743587 Chr.20: 11919618-11919618 on Build GRCh38
rs770398705 Chr.20: 11919860-11919860 on Build GRCh38
rs754951781 Chr.20: 11919769-11919769 on Build GRCh38
rs112611913 Chr.20: 11919943-11919948 on Build GRCh38
rs772388000 Chr.20: 11919849-11919849 on Build GRCh38
rs749857936 Chr.20: 11919680-11919680 on Build GRCh38
rs375919920 Chr.20: 11919753-11919753 on Build GRCh38
rs776127166 Chr.20: 11919673-11919673 on Build GRCh38
rs377208664 Chr.20: 11919601-11919601 on Build GRCh38
rs533638019 Chr.20: 11919943-11919943 on Build GRCh38
rs571169761 Chr.20: 11919615-11919615 on Build GRCh38
rs780319807 Chr.20: 11919814-11919814 on Build GRCh38
rs969447929 Chr.20: 11919928-11919928 on Build GRCh38
rs918453558 Chr.20: 11920002-11920002 on Build GRCh38
rs992447027 Chr.20: 11919877-11919877 on Build GRCh38
rs150544423 Chr.20: 11919829-11919829 on Build GRCh38
rs933890536 Chr.20: 11920037-11920037 on Build GRCh38
rs139514711 Chr.20: 11919793-11919793 on Build GRCh38
rs889449360 Chr.20: 11920047-11920047 on Build GRCh38
rs888271479 Chr.20: 11919670-11919670 on Build GRCh38
rs557622634 Chr.20: 11919578-11919578 on Build GRCh38
rs779301739 Chr.20: 11919839-11919839 on Build GRCh38
rs768604188 Chr.20: 11919901-11919901 on Build GRCh38
rs774392840 Chr.20: 11919993-11919993 on Build GRCh38
rs34603044 Chr.20: 11919999-11920000 on Build GRCh38
rs746149487 Chr.20: 11919796-11919796 on Build GRCh38
rs549059867 Chr.20: 11920014-11920014 on Build GRCh38
rs763843905 Chr.20: 11919880-11919880 on Build GRCh38
rs377229883 Chr.20: 11919822-11919822 on Build GRCh38
rs116150182 Chr.20: 11919850-11919850 on Build GRCh38
rs948522060 Chr.20: 11919617-11919617 on Build GRCh38
rs770066602 Chr.20: 11919672-11919672 on Build GRCh38
rs768869968 Chr.20: 11919696-11919696 on Build GRCh38
rs948700515 Chr.20: 11919950-11919950 on Build GRCh38
rs141676257 Chr.20: 11919794-11919794 on Build GRCh38
rs1051057467 Chr.20: 11920038-11920038 on Build GRCh38
rs760177669 Chr.20: 11919750-11919750 on Build GRCh38
rs529007288 Chr.20: 11919995-11919995 on Build GRCh38
rs915781081 Chr.20: 11919949-11919949 on Build GRCh38
rs540248206 Chr.20: 11919944-11919944 on Build GRCh38
rs992580981 Chr.20: 11919599-11919599 on Build GRCh38
rs779711506 Chr.20: 11919888-11919890 on Build GRCh38
rs767141748 Chr.20: 11919711-11919711 on Build GRCh38
rs909301888 Chr.20: 11919979-11919979 on Build GRCh38
rs916554724 Chr.20: 11919881-11919881 on Build GRCh38
rs753757500 Chr.20: 11919762-11919762 on Build GRCh38
rs941160759 Chr.20: 11919989-11919989 on Build GRCh38
rs765151202 Chr.20: 11919869-11919869 on Build GRCh38
rs1049163779 Chr.20: 11919627-11919627 on Build GRCh38
rs552923117 Chr.20: 11919707-11919707 on Build GRCh38
rs752475790 Chr.20: 11919792-11919792 on Build GRCh38
rs560078067 Chr.20: 11919951-11919951 on Build GRCh38
rs771245955 Chr.20: 11919878-11919879 on Build GRCh38
rs564771152 Chr.20: 11919862-11919862 on Build GRCh38
rs774739952 Chr.20: 11919704-11919704 on Build GRCh38
rs759057386 Chr.20: 11919867-11919867 on Build GRCh38
rs762793985 Chr.20: 11919878-11919878 on Build GRCh38
rs539452687 Chr.20: 11919631-11919631 on Build GRCh38
rs749413025 Chr.20: 11919826-11919826 on Build GRCh38
rs752640599 Chr.20: 11919871-11919871 on Build GRCh38
rs772744537 Chr.20: 11919713-11919713 on Build GRCh38
rs765107401 Chr.20: 11919786-11919786 on Build GRCh38
rs562583790 Chr.20: 11920048-11920048 on Build GRCh38
rs1031231187 Chr.20: 11919854-11919854 on Build GRCh38
rs386812549 Chr.20: 11919943-11919951 on Build GRCh38

View Full Product Details