Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 11922845-11923394 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD3
Entrez Gene ID
22903
Gene Name
BTB domain containing 3
Gene Aliases
dJ742J24.1
UniGene
Hs.709366
Gene Chromosome Location
Chr.20: 11890723-11926595 on Build GRCh38
Transcripts
NM_001282550.1 NM_001282551.1 NM_001282552.2 NM_001282554.1 NM_014962.3 NM_181443.3 XM_017027728.1 XM_017027729.1 XM_017027730.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1024513 c.919G>T p.E307* Chr.20: 11923016-11923016 on Build GRCh38
5391562 c.820C>T p.R274C Chr.20: 11922917-11922917 on Build GRCh38
1615323 c.1103A>T p.K368M Chr.20: 11923200-11923200 on Build GRCh38
6159219 c.1174G>C p.G392R Chr.20: 11923271-11923271 on Build GRCh38
1024515 c.984T>C p.D328D Chr.20: 11923081-11923081 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145986671 Chr.20: 11923191-11923191 on Build GRCh38
rs748681909 Chr.20: 11922988-11922988 on Build GRCh38
rs368565346 Chr.20: 11923269-11923269 on Build GRCh38
rs754208193 Chr.20: 11923299-11923299 on Build GRCh38
rs375257903 Chr.20: 11922884-11922884 on Build GRCh38
rs770597409 Chr.20: 11923224-11923224 on Build GRCh38
rs55860534 Chr.20: 11922883-11922883 on Build GRCh38
rs746273206 Chr.20: 11922950-11922950 on Build GRCh38
rs137886713 Chr.20: 11923164-11923164 on Build GRCh38
rs748246689 Chr.20: 11923123-11923123 on Build GRCh38
rs768807931 Chr.20: 11923248-11923248 on Build GRCh38
rs200448067 Chr.20: 11923025-11923025 on Build GRCh38
rs757592297 Chr.20: 11922928-11922928 on Build GRCh38
rs957592663 Chr.20: 11923044-11923044 on Build GRCh38
rs760934319 Chr.20: 11923172-11923172 on Build GRCh38
rs919178857 Chr.20: 11923026-11923026 on Build GRCh38
rs757820195 Chr.20: 11923351-11923351 on Build GRCh38
rs772251588 Chr.20: 11923264-11923264 on Build GRCh38
rs749150016 Chr.20: 11922922-11922923 on Build GRCh38
rs763696902 Chr.20: 11923358-11923358 on Build GRCh38
rs535169870 Chr.20: 11923236-11923236 on Build GRCh38
rs755672043 Chr.20: 11922981-11922981 on Build GRCh38
rs779135038 Chr.20: 11923070-11923070 on Build GRCh38
rs775548454 Chr.20: 11923125-11923125 on Build GRCh38
rs867698326 Chr.20: 11923059-11923059 on Build GRCh38
rs780999560 Chr.20: 11923231-11923231 on Build GRCh38
rs143653244 Chr.20: 11923111-11923111 on Build GRCh38
rs570256339 Chr.20: 11923159-11923159 on Build GRCh38
rs747696693 Chr.20: 11923105-11923105 on Build GRCh38
rs747013679 Chr.20: 11923222-11923222 on Build GRCh38
rs761695813 Chr.20: 11922876-11922876 on Build GRCh38
rs765305999 Chr.20: 11923028-11923028 on Build GRCh38
rs754515054 Chr.20: 11923079-11923079 on Build GRCh38
rs530038210 Chr.20: 11922918-11922918 on Build GRCh38
rs775846113 Chr.20: 11923029-11923029 on Build GRCh38
rs140411069 Chr.20: 11923135-11923135 on Build GRCh38
rs753584721 Chr.20: 11923078-11923078 on Build GRCh38
rs764920317 Chr.20: 11923309-11923309 on Build GRCh38
rs754641290 Chr.20: 11923197-11923197 on Build GRCh38
rs752200492 Chr.20: 11923211-11923211 on Build GRCh38
rs759996580 Chr.20: 11923304-11923304 on Build GRCh38
rs772844774 Chr.20: 11923013-11923013 on Build GRCh38
rs764578522 Chr.20: 11922922-11922922 on Build GRCh38
rs768694764 Chr.20: 11923207-11923208 on Build GRCh38
rs774436479 Chr.20: 11922866-11922866 on Build GRCh38
rs201830553 Chr.20: 11923308-11923308 on Build GRCh38
rs868536529 Chr.20: 11923045-11923045 on Build GRCh38
rs774418009 Chr.20: 11923005-11923005 on Build GRCh38
rs779571712 Chr.20: 11923074-11923074 on Build GRCh38
rs367831173 Chr.20: 11923237-11923237 on Build GRCh38
rs752452079 Chr.20: 11923350-11923350 on Build GRCh38
rs761877995 Chr.20: 11923262-11923262 on Build GRCh38
rs774383036 Chr.20: 11923210-11923211 on Build GRCh38
rs142386919 Chr.20: 11923180-11923180 on Build GRCh38
rs148938861 Chr.20: 11923008-11923008 on Build GRCh38
rs368296751 Chr.20: 11923108-11923108 on Build GRCh38
rs752028767 Chr.20: 11923039-11923039 on Build GRCh38
rs768090540 Chr.20: 11923054-11923054 on Build GRCh38
rs35364034 Chr.20: 11923006-11923006 on Build GRCh38
rs766829626 Chr.20: 11923285-11923285 on Build GRCh38
rs764204510 Chr.20: 11923034-11923034 on Build GRCh38
rs138797432 Chr.20: 11923217-11923217 on Build GRCh38
rs996360240 Chr.20: 11922964-11922964 on Build GRCh38
rs751007170 Chr.20: 11923168-11923168 on Build GRCh38
rs750695150 Chr.20: 11923067-11923067 on Build GRCh38
rs779772020 Chr.20: 11922983-11922983 on Build GRCh38
rs768347962 Chr.20: 11922991-11922991 on Build GRCh38
rs777556357 Chr.20: 11923218-11923218 on Build GRCh38
rs930540638 Chr.20: 11923268-11923268 on Build GRCh38
rs150389358 Chr.20: 11923160-11923160 on Build GRCh38
rs763002395 Chr.20: 11923030-11923030 on Build GRCh38
rs964842026 Chr.20: 11922948-11922948 on Build GRCh38
rs781576058 Chr.20: 11922946-11922946 on Build GRCh38
rs142793480 Chr.20: 11923371-11923371 on Build GRCh38
rs201543769 Chr.20: 11923258-11923258 on Build GRCh38
rs778733657 Chr.20: 11923210-11923210 on Build GRCh38
rs752114393 Chr.20: 11922926-11922926 on Build GRCh38
rs547995041 Chr.20: 11923189-11923189 on Build GRCh38
rs778611018 Chr.20: 11923081-11923081 on Build GRCh38
rs765529359 Chr.20: 11922914-11922914 on Build GRCh38
rs761873657 Chr.20: 11923275-11923283 on Build GRCh38
rs560899008 Chr.20: 11922917-11922917 on Build GRCh38
rs199603777 Chr.20: 11923049-11923049 on Build GRCh38
rs768897473 Chr.20: 11923144-11923144 on Build GRCh38

View Full Product Details