Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 103319516-103320064 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC2
Entrez Gene ID
27000
Gene Name
DnaJ heat shock protein family (Hsp40) member C2
Gene Aliases
MPHOSPH11, MPP11, ZRF1, ZUO1
UniGene
Hs.558476
Gene Chromosome Location
Chr.7: 103312474-103345362 on Build GRCh38
Transcripts
NM_001129887.1 NM_014377.1 XM_005250269.2 XM_011516028.2 XM_011516029.2 XM_011516030.2 XM_017011968.1 XM_005250717.3 XM_006716181.3

Gene Symbol
PMPCB
Entrez Gene ID
9512
Gene Name
peptidase, mitochondrial processing beta subunit
Gene Aliases
Beta-MPP, MPP11, MPPB, MPPP52, P-52
UniGene
Hs.184211
Gene Chromosome Location
Chr.7: 103297412-103329902 on Build GRCh38
Transcripts
NM_001129887.1 NM_014377.1 XM_005250269.2 XM_011516028.2 XM_011516029.2 XM_011516030.2 XM_017011968.1 XM_005250717.3 XM_006716181.3

SNP Detail

SNP ID Chromosome Location
rs370268421 Chr.7: 103319853-103319853 on Build GRCh38
rs373699568 Chr.7: 103319875-103319875 on Build GRCh38
rs965934815 Chr.7: 103319990-103319990 on Build GRCh38
rs761681108 Chr.7: 103319877-103319877 on Build GRCh38
rs952080649 Chr.7: 103320013-103320013 on Build GRCh38
rs753697535 Chr.7: 103319730-103319730 on Build GRCh38
rs975734560 Chr.7: 103319671-103319671 on Build GRCh38
rs1044183373 Chr.7: 103319928-103319928 on Build GRCh38
rs1032604057 Chr.7: 103319952-103319952 on Build GRCh38
rs903667858 Chr.7: 103319971-103319971 on Build GRCh38
rs752395905 Chr.7: 103319717-103319717 on Build GRCh38
rs374725378 Chr.7: 103319686-103319686 on Build GRCh38
rs773634289 Chr.7: 103319635-103319635 on Build GRCh38
rs748748450 Chr.7: 103319617-103319617 on Build GRCh38
rs770053453 Chr.7: 103319631-103319631 on Build GRCh38
rs974963529 Chr.7: 103319991-103319991 on Build GRCh38
rs955926545 Chr.7: 103319579-103319579 on Build GRCh38
rs559940336 Chr.7: 103319982-103319982 on Build GRCh38
rs777361207 Chr.7: 103319861-103319861 on Build GRCh38
rs760471331 Chr.7: 103319862-103319862 on Build GRCh38
rs527526626 Chr.7: 103319927-103319927 on Build GRCh38
rs955957755 Chr.7: 103319957-103319957 on Build GRCh38
rs921997739 Chr.7: 103319734-103319734 on Build GRCh38
rs201791551 Chr.7: 103319771-103319771 on Build GRCh38
rs552780373 Chr.7: 103319930-103319930 on Build GRCh38
rs781529486 Chr.7: 103319605-103319605 on Build GRCh38
rs758024172 Chr.7: 103319599-103319599 on Build GRCh38
rs182597576 Chr.7: 103319578-103319578 on Build GRCh38
rs775643727 Chr.7: 103319859-103319859 on Build GRCh38
rs999385201 Chr.7: 103319983-103319983 on Build GRCh38
rs766458672 Chr.7: 103319685-103319685 on Build GRCh38
rs75518967 Chr.7: 103319766-103319766 on Build GRCh38
rs749687683 Chr.7: 103319818-103319818 on Build GRCh38
rs754876265 Chr.7: 103319692-103319694 on Build GRCh38
rs374873868 Chr.7: 103319785-103319785 on Build GRCh38
rs752717085 Chr.7: 103319572-103319572 on Build GRCh38
rs759578666 Chr.7: 103319683-103319683 on Build GRCh38
rs765182154 Chr.7: 103319887-103319887 on Build GRCh38
rs746142800 Chr.7: 103319843-103319843 on Build GRCh38
rs201350713 Chr.7: 103319587-103319587 on Build GRCh38
rs922030299 Chr.7: 103320000-103320000 on Build GRCh38
rs779682332 Chr.7: 103319799-103319799 on Build GRCh38
rs377693338 Chr.7: 103319589-103319589 on Build GRCh38
rs557018963 Chr.7: 103319814-103319814 on Build GRCh38
rs778506913 Chr.7: 103319596-103319596 on Build GRCh38
rs753680898 Chr.7: 103319685-103319687 on Build GRCh38
rs200619898 Chr.7: 103319823-103319823 on Build GRCh38
rs756662289 Chr.7: 103319810-103319810 on Build GRCh38
rs767681306 Chr.7: 103319688-103319688 on Build GRCh38
rs755491441 Chr.7: 103319548-103319548 on Build GRCh38
rs763729088 Chr.7: 103319709-103319709 on Build GRCh38
rs757803600 Chr.7: 103319821-103319823 on Build GRCh38
rs778369168 Chr.7: 103319707-103319708 on Build GRCh38
rs776525510 Chr.7: 103319876-103319876 on Build GRCh38
rs753082361 Chr.7: 103319801-103319801 on Build GRCh38
rs749456723 Chr.7: 103319654-103319654 on Build GRCh38
rs374877873 Chr.7: 103319917-103319917 on Build GRCh38
rs557810300 Chr.7: 103319929-103319929 on Build GRCh38
rs878856228 Chr.7: 103320025-103320025 on Build GRCh38
rs148077765 Chr.7: 103320021-103320021 on Build GRCh38
rs377679257 Chr.7: 103319735-103319735 on Build GRCh38
rs369916864 Chr.7: 103319660-103319660 on Build GRCh38
rs771322302 Chr.7: 103319821-103319821 on Build GRCh38
rs374132344 Chr.7: 103319691-103319691 on Build GRCh38
rs757968839 Chr.7: 103319772-103319772 on Build GRCh38
rs538091160 Chr.7: 103319684-103319685 on Build GRCh38
rs1055803857 Chr.7: 103319889-103319889 on Build GRCh38
rs760732650 Chr.7: 103319693-103319693 on Build GRCh38
rs1019270292 Chr.7: 103319970-103319970 on Build GRCh38
rs747655475 Chr.7: 103319669-103319669 on Build GRCh38
rs199525138 Chr.7: 103319792-103319792 on Build GRCh38
rs764953269 Chr.7: 103319739-103319739 on Build GRCh38
rs779456178 Chr.7: 103319637-103319637 on Build GRCh38
rs767383519 Chr.7: 103319568-103319568 on Build GRCh38
rs143667678 Chr.7: 103319966-103319966 on Build GRCh38
rs894521266 Chr.7: 103319905-103319905 on Build GRCh38
rs755839389 Chr.7: 103319577-103319577 on Build GRCh38
rs901609370 Chr.7: 103320002-103320002 on Build GRCh38
rs540342256 Chr.7: 103319967-103319967 on Build GRCh38
rs908137685 Chr.7: 103319779-103319779 on Build GRCh38

View Full Product Details