Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 136344920-136345147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRG4
Entrez Gene ID
139378
Gene Name
adhesion G protein-coupled receptor G4
Gene Aliases
GPR112, PGR17, RP1-299I16
UniGene
Hs.381354
Gene Chromosome Location
Chr.X: 136300963-136416888 on Build GRCh38
Transcripts
NM_153834.3 XM_011531269.2 XM_011531271.2

SNP Detail

SNP ID Chromosome Location
rs758504607 Chr.X: 136345012-136345012 on Build GRCh38
rs898147415 Chr.X: 136345054-136345054 on Build GRCh38
rs771623890 Chr.X: 136345049-136345049 on Build GRCh38
rs914573290 Chr.X: 136344972-136344972 on Build GRCh38
rs140366775 Chr.X: 136344988-136344988 on Build GRCh38
rs149725423 Chr.X: 136345036-136345036 on Build GRCh38
rs750941890 Chr.X: 136344964-136344964 on Build GRCh38
rs901949385 Chr.X: 136345007-136345007 on Build GRCh38
rs757913874 Chr.X: 136345079-136345079 on Build GRCh38
rs777648603 Chr.X: 136345021-136345021 on Build GRCh38
rs147597943 Chr.X: 136344974-136344974 on Build GRCh38
rs369394930 Chr.X: 136344959-136344959 on Build GRCh38
rs779724533 Chr.X: 136345096-136345096 on Build GRCh38
rs893538625 Chr.X: 136345008-136345008 on Build GRCh38
rs752921941 Chr.X: 136345122-136345122 on Build GRCh38
rs185759057 Chr.X: 136345119-136345119 on Build GRCh38
rs1015232680 Chr.X: 136345104-136345104 on Build GRCh38
rs769610298 Chr.X: 136345031-136345031 on Build GRCh38
rs765548885 Chr.X: 136345120-136345120 on Build GRCh38
rs180765843 Chr.X: 136344991-136344991 on Build GRCh38
rs763813411 Chr.X: 136345082-136345082 on Build GRCh38
rs772583051 Chr.X: 136345058-136345058 on Build GRCh38
rs761487047 Chr.X: 136345046-136345046 on Build GRCh38
rs745835572 Chr.X: 136345023-136345023 on Build GRCh38
rs761308517 Chr.X: 136345071-136345071 on Build GRCh38
rs375616928 Chr.X: 136345077-136345077 on Build GRCh38
rs752868855 Chr.X: 136344992-136344992 on Build GRCh38
rs1050888314 Chr.X: 136345027-136345027 on Build GRCh38
rs777104187 Chr.X: 136345116-136345116 on Build GRCh38
rs371997210 Chr.X: 136345017-136345017 on Build GRCh38
rs933439915 Chr.X: 136345022-136345022 on Build GRCh38
rs749064564 Chr.X: 136345035-136345035 on Build GRCh38
rs761028829 Chr.X: 136344970-136344970 on Build GRCh38
rs754146292 Chr.X: 136344985-136344985 on Build GRCh38
rs145542316 Chr.X: 136345126-136345126 on Build GRCh38
rs1016901107 Chr.X: 136345040-136345040 on Build GRCh38

View Full Product Details