Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 11922482-11922755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD3
Entrez Gene ID
22903
Gene Name
BTB domain containing 3
Gene Aliases
dJ742J24.1
UniGene
Hs.709366
Gene Chromosome Location
Chr.20: 11890723-11926595 on Build GRCh38
Transcripts
NM_001282550.1 NM_001282551.1 NM_001282552.2 NM_001282554.1 NM_014962.3 NM_181443.3 XM_017027728.1 XM_017027729.1 XM_017027730.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
234455 c.645C>T p.F215F Chr.20: 11922742-11922742 on Build GRCh38
3840407 c.620A>C p.H207P Chr.20: 11922717-11922717 on Build GRCh38
443455 c.546T>C p.Y182Y Chr.20: 11922643-11922643 on Build GRCh38
1024507 c.565G>A p.A189T Chr.20: 11922662-11922662 on Build GRCh38
5587308 c.615C>T p.V205V Chr.20: 11922712-11922712 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776267778 Chr.20: 11922643-11922643 on Build GRCh38
rs755076085 Chr.20: 11922722-11922722 on Build GRCh38
rs140980126 Chr.20: 11922516-11922516 on Build GRCh38
rs747407766 Chr.20: 11922631-11922631 on Build GRCh38
rs780446894 Chr.20: 11922591-11922591 on Build GRCh38
rs750640654 Chr.20: 11922586-11922586 on Build GRCh38
rs759078145 Chr.20: 11922649-11922649 on Build GRCh38
rs575631924 Chr.20: 11922637-11922637 on Build GRCh38
rs146595270 Chr.20: 11922703-11922703 on Build GRCh38
rs769242030 Chr.20: 11922650-11922650 on Build GRCh38
rs960264509 Chr.20: 11922518-11922518 on Build GRCh38
rs778912914 Chr.20: 11922727-11922727 on Build GRCh38
rs754181251 Chr.20: 11922712-11922712 on Build GRCh38
rs779471241 Chr.20: 11922609-11922609 on Build GRCh38
rs373656470 Chr.20: 11922605-11922605 on Build GRCh38
rs967908555 Chr.20: 11922571-11922571 on Build GRCh38
rs748665437 Chr.20: 11922613-11922613 on Build GRCh38
rs1020509150 Chr.20: 11922569-11922569 on Build GRCh38
rs181451510 Chr.20: 11922614-11922614 on Build GRCh38
rs989239401 Chr.20: 11922521-11922521 on Build GRCh38
rs373540138 Chr.20: 11922638-11922638 on Build GRCh38
rs756362962 Chr.20: 11922588-11922588 on Build GRCh38
rs751352460 Chr.20: 11922685-11922685 on Build GRCh38
rs913257847 Chr.20: 11922531-11922531 on Build GRCh38
rs762837488 Chr.20: 11922658-11922658 on Build GRCh38
rs370980413 Chr.20: 11922652-11922652 on Build GRCh38
rs755017888 Chr.20: 11922608-11922608 on Build GRCh38
rs376461626 Chr.20: 11922662-11922662 on Build GRCh38
rs761716128 Chr.20: 11922695-11922695 on Build GRCh38
rs140075905 Chr.20: 11922620-11922620 on Build GRCh38
rs143395509 Chr.20: 11922555-11922555 on Build GRCh38

View Full Product Details