Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 14850458-14850731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCB
Entrez Gene ID
2187
Gene Name
Fanconi anemia complementation group B
Gene Aliases
FA2, FAAP90, FAAP95, FAB, FACB
UniGene
Hs.554740
Gene Chromosome Location
Chr.X: 14690863-14873255 on Build GRCh38
Transcripts
NM_001018113.2 NM_001324162.1 NM_152633.3 XM_011545470.2 XM_017029355.1 XM_017029356.1

SNP Detail

SNP ID Chromosome Location
rs757930604 Chr.X: 14850491-14850491 on Build GRCh38
rs140773512 Chr.X: 14850628-14850628 on Build GRCh38
rs993307430 Chr.X: 14850514-14850514 on Build GRCh38
rs754738590 Chr.X: 14850694-14850694 on Build GRCh38
rs970019905 Chr.X: 14850650-14850650 on Build GRCh38
rs143976596 Chr.X: 14850608-14850608 on Build GRCh38
rs778796466 Chr.X: 14850579-14850579 on Build GRCh38
rs140198444 Chr.X: 14850559-14850559 on Build GRCh38
rs374559250 Chr.X: 14850599-14850599 on Build GRCh38
rs761286423 Chr.X: 14850647-14850647 on Build GRCh38
rs764817410 Chr.X: 14850652-14850652 on Build GRCh38
rs781087988 Chr.X: 14850546-14850546 on Build GRCh38
rs746103110 Chr.X: 14850595-14850595 on Build GRCh38
rs766440560 Chr.X: 14850690-14850693 on Build GRCh38
rs754933644 Chr.X: 14850518-14850518 on Build GRCh38
rs750596911 Chr.X: 14850676-14850677 on Build GRCh38
rs562994158 Chr.X: 14850691-14850691 on Build GRCh38
rs368679125 Chr.X: 14850670-14850670 on Build GRCh38
rs146438837 Chr.X: 14850611-14850611 on Build GRCh38
rs867778700 Chr.X: 14850659-14850659 on Build GRCh38
rs372644335 Chr.X: 14850575-14850575 on Build GRCh38
rs2905223 Chr.X: 14850684-14850684 on Build GRCh38
rs202067682 Chr.X: 14850677-14850677 on Build GRCh38
rs746768201 Chr.X: 14850508-14850508 on Build GRCh38
rs935080919 Chr.X: 14850505-14850505 on Build GRCh38
rs781260236 Chr.X: 14850656-14850656 on Build GRCh38
rs149695930 Chr.X: 14850630-14850630 on Build GRCh38
rs781062442 Chr.X: 14850696-14850696 on Build GRCh38
rs777190282 Chr.X: 14850643-14850643 on Build GRCh38
rs199510538 Chr.X: 14850507-14850507 on Build GRCh38
rs201873039 Chr.X: 14850489-14850489 on Build GRCh38

View Full Product Details