Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 75578903-75579160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLVCR2
Entrez Gene ID
55640
Gene Name
feline leukemia virus subgroup C cellular receptor family member 2
Gene Aliases
C14orf58, CCT, EPV, FLVCRL14q, MFSD7C, PVHH
UniGene
Hs.509966
Gene Chromosome Location
Chr.14: 75578597-75648169 on Build GRCh38
Transcripts
NM_017791.2 NR_110552.1

Gene Symbol
LOC102724153
Entrez Gene ID
102724153
Gene Name
uncharacterized LOC102724153
Gene Aliases
-
UniGene
Hs.615289
Gene Chromosome Location
Chr.14: 75574901-75579588 on Build GRCh38
Transcripts
NM_017791.2 NR_110552.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2138653 c.102G>C p.S34S Chr.14: 75579074-75579074 on Build GRCh38
2138654 c.103G>A p.V35I Chr.14: 75579075-75579075 on Build GRCh38
7129048 c.83C>G p.S28W Chr.14: 75579055-75579055 on Build GRCh38
2138655 c.106C>A p.H36N Chr.14: 75579078-75579078 on Build GRCh38
7129049 c.137C>G p.S46C Chr.14: 75579109-75579109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369546881 Chr.14: 75579117-75579117 on Build GRCh38
rs367807214 Chr.14: 75578970-75578970 on Build GRCh38
rs778276591 Chr.14: 75579097-75579114 on Build GRCh38
rs762529776 Chr.14: 75579013-75579013 on Build GRCh38
rs751059938 Chr.14: 75579099-75579099 on Build GRCh38
rs773193566 Chr.14: 75579098-75579099 on Build GRCh38
rs763882900 Chr.14: 75579081-75579098 on Build GRCh38
rs372243784 Chr.14: 75579136-75579136 on Build GRCh38
rs371981338 Chr.14: 75579036-75579036 on Build GRCh38
rs749569624 Chr.14: 75579045-75579045 on Build GRCh38
rs143304751 Chr.14: 75579075-75579075 on Build GRCh38
rs761432705 Chr.14: 75579101-75579101 on Build GRCh38
rs977013723 Chr.14: 75579140-75579140 on Build GRCh38
rs753823415 Chr.14: 75579080-75579081 on Build GRCh38
rs781206373 Chr.14: 75579029-75579029 on Build GRCh38
rs1035510024 Chr.14: 75579063-75579063 on Build GRCh38
rs148295074 Chr.14: 75579124-75579124 on Build GRCh38
rs1013248578 Chr.14: 75579105-75579105 on Build GRCh38
rs773848595 Chr.14: 75579032-75579032 on Build GRCh38
rs376955487 Chr.14: 75579107-75579107 on Build GRCh38
rs2287015 Chr.14: 75579019-75579019 on Build GRCh38
rs748229824 Chr.14: 75579065-75579065 on Build GRCh38
rs548569935 Chr.14: 75579045-75579062 on Build GRCh38
rs751540374 Chr.14: 75579022-75579022 on Build GRCh38
rs752434981 Chr.14: 75579003-75579003 on Build GRCh38
rs772508772 Chr.14: 75579069-75579069 on Build GRCh38
rs183899065 Chr.14: 75578942-75578942 on Build GRCh38
rs762762330 Chr.14: 75579079-75579079 on Build GRCh38
rs779028741 Chr.14: 75579056-75579056 on Build GRCh38
rs750140841 Chr.14: 75579030-75579030 on Build GRCh38
rs756068385 Chr.14: 75579044-75579045 on Build GRCh38
rs778813108 Chr.14: 75579105-75579140 on Build GRCh38
rs927307658 Chr.14: 75578992-75578992 on Build GRCh38
rs773791424 Chr.14: 75579074-75579074 on Build GRCh38
rs768707435 Chr.14: 75579050-75579050 on Build GRCh38
rs980065536 Chr.14: 75578989-75578989 on Build GRCh38
rs764747260 Chr.14: 75579002-75579002 on Build GRCh38
rs199789502 Chr.14: 75579128-75579128 on Build GRCh38
rs780104839 Chr.14: 75579038-75579038 on Build GRCh38
rs763845250 Chr.14: 75579082-75579082 on Build GRCh38
rs868407513 Chr.14: 75579130-75579130 on Build GRCh38
rs761114230 Chr.14: 75578980-75578980 on Build GRCh38
rs757198109 Chr.14: 75579025-75579025 on Build GRCh38

View Full Product Details