Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 492195-492420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK2A1
Entrez Gene ID
1457
Gene Name
casein kinase 2 alpha 1
Gene Aliases
CK2A1, CKII, CSNK2A3
UniGene
Hs.644056
Gene Chromosome Location
Chr.20: 482694-543838 on Build GRCh38
Transcripts
NM_001895.3 NM_177559.2 NM_177560.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4974556 c.547C>T p.H183Y Chr.20: 492328-492328 on Build GRCh38
7106395 c.523G>C p.D175H Chr.20: 492352-492352 on Build GRCh38
3295369 c.584G>A p.R195Q Chr.20: 492291-492291 on Build GRCh38
5903245 c.581C>T p.S194F Chr.20: 492294-492294 on Build GRCh38
1027637 c.599C>T p.P200L Chr.20: 492276-492276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776269968 Chr.20: 492394-492394 on Build GRCh38
rs772093878 Chr.20: 492356-492356 on Build GRCh38
rs770574791 Chr.20: 492393-492393 on Build GRCh38
rs1011711423 Chr.20: 492380-492380 on Build GRCh38
rs759151227 Chr.20: 492395-492395 on Build GRCh38
rs780238300 Chr.20: 492308-492308 on Build GRCh38
rs751999830 Chr.20: 492247-492247 on Build GRCh38
rs372557928 Chr.20: 492329-492329 on Build GRCh38
rs772991353 Chr.20: 492373-492373 on Build GRCh38
rs919408183 Chr.20: 492272-492272 on Build GRCh38
rs746026143 Chr.20: 492305-492305 on Build GRCh38
rs869312848 Chr.20: 492351-492351 on Build GRCh38
rs748116533 Chr.20: 492312-492312 on Build GRCh38
rs547574228 Chr.20: 492302-492302 on Build GRCh38
rs869312840 Chr.20: 492282-492282 on Build GRCh38
rs375003251 Chr.20: 492225-492225 on Build GRCh38
rs910590145 Chr.20: 492384-492384 on Build GRCh38
rs1034583315 Chr.20: 492292-492292 on Build GRCh38
rs369232772 Chr.20: 492269-492269 on Build GRCh38
rs769996282 Chr.20: 492306-492306 on Build GRCh38
rs760584646 Chr.20: 492386-492386 on Build GRCh38
rs376824398 Chr.20: 492385-492385 on Build GRCh38

View Full Product Details