Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219221843-219222086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG9A
Entrez Gene ID
79065
Gene Name
autophagy related 9A
Gene Aliases
APG9L1, MGD3208, mATG9
UniGene
Hs.323363
Gene Chromosome Location
Chr.2: 219219380-219229688 on Build GRCh38
Transcripts
NM_001077198.2 NM_024085.4 NR_104255.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6378522 c.2120G>A p.S707N Chr.2: 219222075-219222075 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371204401 Chr.2: 219222040-219222040 on Build GRCh38
rs771414973 Chr.2: 219221994-219221994 on Build GRCh38
rs369753732 Chr.2: 219222060-219222060 on Build GRCh38
rs985515140 Chr.2: 219221921-219221921 on Build GRCh38
rs757007567 Chr.2: 219222029-219222030 on Build GRCh38
rs373665258 Chr.2: 219221926-219221926 on Build GRCh38
rs929154287 Chr.2: 219221992-219221992 on Build GRCh38
rs780952055 Chr.2: 219222024-219222024 on Build GRCh38
rs367762210 Chr.2: 219222035-219222035 on Build GRCh38
rs753599149 Chr.2: 219222028-219222031 on Build GRCh38
rs897059802 Chr.2: 219221880-219221880 on Build GRCh38
rs998410874 Chr.2: 219221881-219221881 on Build GRCh38
rs1032690988 Chr.2: 219221980-219221980 on Build GRCh38
rs1014076268 Chr.2: 219221883-219221883 on Build GRCh38
rs969834838 Chr.2: 219221927-219221927 on Build GRCh38
rs553160066 Chr.2: 219221993-219221993 on Build GRCh38
rs973229542 Chr.2: 219221983-219221983 on Build GRCh38
rs750258904 Chr.2: 219222027-219222027 on Build GRCh38
rs909947953 Chr.2: 219221937-219221937 on Build GRCh38
rs144521674 Chr.2: 219221915-219221915 on Build GRCh38
rs1001656227 Chr.2: 219221966-219221966 on Build GRCh38
rs760660779 Chr.2: 219222025-219222026 on Build GRCh38
rs763702485 Chr.2: 219222028-219222030 on Build GRCh38
rs376837244 Chr.2: 219222008-219222008 on Build GRCh38
rs760360016 Chr.2: 219222023-219222024 on Build GRCh38
rs77197458 Chr.2: 219222049-219222049 on Build GRCh38
rs200215453 Chr.2: 219222009-219222009 on Build GRCh38
rs919067292 Chr.2: 219221985-219221985 on Build GRCh38
rs768200397 Chr.2: 219222055-219222055 on Build GRCh38
rs376745930 Chr.2: 219222041-219222041 on Build GRCh38
rs765784423 Chr.2: 219221995-219221995 on Build GRCh38
rs948382997 Chr.2: 219222043-219222043 on Build GRCh38
rs779689308 Chr.2: 219222039-219222039 on Build GRCh38

View Full Product Details