Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 61703811-61704084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf64
Entrez Gene ID
285668
Gene Name
chromosome 5 open reading frame 64
Gene Aliases
-
UniGene
Hs.683866
Gene Chromosome Location
Chr.5: 61637308-61759165 on Build GRCh38
Transcripts
NM_173667.3 NR_126523.1 NR_126524.1 NR_126525.1 XM_017009378.1 XM_017009379.1 XM_017009380.1 XM_017009381.1 XM_017009382.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4625579 c.327G>A p.M109I Chr.5: 61703967-61703967 on Build GRCh38
7269627 c.276C>T p.N92N Chr.5: 61703916-61703916 on Build GRCh38
1438081 c.301C>T p.R101W Chr.5: 61703941-61703941 on Build GRCh38
212554 c.244C>T p.P82S Chr.5: 61703884-61703884 on Build GRCh38
3393626 c.243T>A p.H81Q Chr.5: 61703883-61703883 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377529422 Chr.5: 61704029-61704029 on Build GRCh38
rs989542381 Chr.5: 61703985-61703985 on Build GRCh38
rs749168375 Chr.5: 61703981-61703981 on Build GRCh38
rs769546277 Chr.5: 61703849-61703849 on Build GRCh38
rs777964184 Chr.5: 61703937-61703937 on Build GRCh38
rs763136427 Chr.5: 61703860-61703860 on Build GRCh38
rs770854766 Chr.5: 61704063-61704063 on Build GRCh38
rs751116297 Chr.5: 61703884-61703885 on Build GRCh38
rs372839363 Chr.5: 61703998-61703998 on Build GRCh38
rs772176937 Chr.5: 61703942-61703942 on Build GRCh38
rs754249807 Chr.5: 61703891-61703891 on Build GRCh38
rs374666655 Chr.5: 61703965-61703965 on Build GRCh38
rs367998263 Chr.5: 61703873-61703873 on Build GRCh38
rs778439357 Chr.5: 61704032-61704035 on Build GRCh38
rs371111757 Chr.5: 61704005-61704005 on Build GRCh38
rs775326846 Chr.5: 61703850-61703850 on Build GRCh38
rs958092064 Chr.5: 61703968-61703968 on Build GRCh38
rs766699921 Chr.5: 61703883-61703883 on Build GRCh38
rs73113137 Chr.5: 61703961-61703961 on Build GRCh38
rs755687941 Chr.5: 61704000-61704000 on Build GRCh38
rs532300347 Chr.5: 61703974-61703974 on Build GRCh38
rs747111859 Chr.5: 61704055-61704055 on Build GRCh38
rs436696 Chr.5: 61703941-61703941 on Build GRCh38
rs376769396 Chr.5: 61703944-61703944 on Build GRCh38
rs778646380 Chr.5: 61704007-61704007 on Build GRCh38
rs577331885 Chr.5: 61703851-61703851 on Build GRCh38
rs761212256 Chr.5: 61703882-61703882 on Build GRCh38
rs779809364 Chr.5: 61703928-61703928 on Build GRCh38
rs760620290 Chr.5: 61703952-61703952 on Build GRCh38
rs767571910 Chr.5: 61703879-61703879 on Build GRCh38
rs756058089 Chr.5: 61703920-61703920 on Build GRCh38
rs147821972 Chr.5: 61703874-61703874 on Build GRCh38
rs773263700 Chr.5: 61704050-61704050 on Build GRCh38
rs774612808 Chr.5: 61703938-61703938 on Build GRCh38
rs745789422 Chr.5: 61703845-61703845 on Build GRCh38
rs767580996 Chr.5: 61703959-61703959 on Build GRCh38
rs762646920 Chr.5: 61703878-61703878 on Build GRCh38
rs773773616 Chr.5: 61703881-61703881 on Build GRCh38
rs751203248 Chr.5: 61703907-61703907 on Build GRCh38
rs772301191 Chr.5: 61704031-61704031 on Build GRCh38
rs755539606 Chr.5: 61703966-61703966 on Build GRCh38
rs373040714 Chr.5: 61704016-61704016 on Build GRCh38
rs562240533 Chr.5: 61703954-61703954 on Build GRCh38
rs369356590 Chr.5: 61703983-61703983 on Build GRCh38
rs781296878 Chr.5: 61703843-61703843 on Build GRCh38
rs368329462 Chr.5: 61703935-61703935 on Build GRCh38
rs745835404 Chr.5: 61703917-61703917 on Build GRCh38
rs76894732 Chr.5: 61704004-61704004 on Build GRCh38
rs375670000 Chr.5: 61703916-61703916 on Build GRCh38
rs76237213 Chr.5: 61703955-61703955 on Build GRCh38
rs563361088 Chr.5: 61703962-61703962 on Build GRCh38
rs755254773 Chr.5: 61703905-61703905 on Build GRCh38
rs749139744 Chr.5: 61703929-61703929 on Build GRCh38
rs754489775 Chr.5: 61703981-61703981 on Build GRCh38
rs374740540 Chr.5: 61703840-61703840 on Build GRCh38
rs141279630 Chr.5: 61703908-61703908 on Build GRCh38

View Full Product Details