Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50177398-50177671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIMA1
Entrez Gene ID
51474
Gene Name
LIM domain and actin binding 1
Gene Aliases
EPLIN, SREBP3
UniGene
Hs.525419
Gene Chromosome Location
Chr.12: 50175780-50283570 on Build GRCh38
Transcripts
NM_001113546.1 NM_001113547.1 NM_001243775.1 NM_016357.4 XM_011538455.2 XM_017019429.1 XM_017019430.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
266824 c.1766G>A p.R589H Chr.12: 50177578-50177578 on Build GRCh38
5422304 c.1773C>A p.F591L Chr.12: 50177571-50177571 on Build GRCh38
7375064 c.1742C>T p.S581F Chr.12: 50177602-50177602 on Build GRCh38
6553266 c.1936G>A p.G646R Chr.12: 50177408-50177408 on Build GRCh38
4474407 c.1916C>T p.A639V Chr.12: 50177428-50177428 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370081421 Chr.12: 50177613-50177613 on Build GRCh38
rs778430209 Chr.12: 50177581-50177581 on Build GRCh38
rs774435355 Chr.12: 50177466-50177467 on Build GRCh38
rs776021559 Chr.12: 50177596-50177598 on Build GRCh38
rs957892686 Chr.12: 50177599-50177599 on Build GRCh38
rs750928636 Chr.12: 50177554-50177554 on Build GRCh38
rs764236862 Chr.12: 50177526-50177526 on Build GRCh38
rs751875072 Chr.12: 50177531-50177531 on Build GRCh38
rs766994071 Chr.12: 50177649-50177649 on Build GRCh38
rs772821365 Chr.12: 50177605-50177605 on Build GRCh38
rs896395477 Chr.12: 50177483-50177483 on Build GRCh38
rs749679966 Chr.12: 50177577-50177577 on Build GRCh38
rs376514299 Chr.12: 50177505-50177505 on Build GRCh38
rs1013469392 Chr.12: 50177569-50177569 on Build GRCh38
rs199825261 Chr.12: 50177627-50177627 on Build GRCh38
rs763113183 Chr.12: 50177525-50177525 on Build GRCh38
rs994395012 Chr.12: 50177470-50177470 on Build GRCh38
rs1025452034 Chr.12: 50177473-50177473 on Build GRCh38
rs766134182 Chr.12: 50177494-50177494 on Build GRCh38
rs564473856 Chr.12: 50177578-50177578 on Build GRCh38
rs1038122775 Chr.12: 50177438-50177438 on Build GRCh38
rs761687778 Chr.12: 50177624-50177624 on Build GRCh38
rs1030866076 Chr.12: 50177576-50177576 on Build GRCh38
rs771683137 Chr.12: 50177466-50177466 on Build GRCh38
rs1014732368 Chr.12: 50177446-50177446 on Build GRCh38
rs781115938 Chr.12: 50177630-50177630 on Build GRCh38
rs771280058 Chr.12: 50177600-50177600 on Build GRCh38
rs963840988 Chr.12: 50177646-50177646 on Build GRCh38
rs756716638 Chr.12: 50177574-50177574 on Build GRCh38
rs771463847 Chr.12: 50177604-50177604 on Build GRCh38
rs544503051 Chr.12: 50177575-50177575 on Build GRCh38
rs1056313137 Chr.12: 50177441-50177441 on Build GRCh38
rs146805564 Chr.12: 50177533-50177533 on Build GRCh38
rs746330915 Chr.12: 50177469-50177470 on Build GRCh38
rs374529313 Chr.12: 50177631-50177631 on Build GRCh38
rs761230915 Chr.12: 50177454-50177454 on Build GRCh38
rs111446133 Chr.12: 50177492-50177492 on Build GRCh38
rs374566973 Chr.12: 50177445-50177445 on Build GRCh38
rs768442080 Chr.12: 50177433-50177433 on Build GRCh38
rs752801137 Chr.12: 50177566-50177566 on Build GRCh38
rs756628778 Chr.12: 50177648-50177648 on Build GRCh38
rs746911119 Chr.12: 50177606-50177606 on Build GRCh38
rs750550657 Chr.12: 50177642-50177642 on Build GRCh38
rs773013856 Chr.12: 50177490-50177490 on Build GRCh38
rs999403806 Chr.12: 50177568-50177568 on Build GRCh38
rs767454940 Chr.12: 50177634-50177634 on Build GRCh38
rs747897491 Chr.12: 50177595-50177595 on Build GRCh38
rs1006679530 Chr.12: 50177435-50177435 on Build GRCh38
rs772533666 Chr.12: 50177523-50177524 on Build GRCh38

View Full Product Details