Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50177612-50177837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIMA1
Entrez Gene ID
51474
Gene Name
LIM domain and actin binding 1
Gene Aliases
EPLIN, SREBP3
UniGene
Hs.525419
Gene Chromosome Location
Chr.12: 50175780-50283570 on Build GRCh38
Transcripts
NM_001113546.1 NM_001113547.1 NM_001243775.1 NM_016357.4 XM_011538455.2 XM_017019429.1 XM_017019430.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6439941 c.1579A>T p.K527* Chr.12: 50177765-50177765 on Build GRCh38
6000253 c.1713C>T p.V571V Chr.12: 50177631-50177631 on Build GRCh38
4467888 c.1509C>T p.V503V Chr.12: 50177835-50177835 on Build GRCh38
7417696 c.1541C>G p.S514C Chr.12: 50177803-50177803 on Build GRCh38
6072793 c.1543C>G p.Q515E Chr.12: 50177801-50177801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963840988 Chr.12: 50177646-50177646 on Build GRCh38
rs186777719 Chr.12: 50177684-50177684 on Build GRCh38
rs980176879 Chr.12: 50177661-50177661 on Build GRCh38
rs910930443 Chr.12: 50177795-50177795 on Build GRCh38
rs910123216 Chr.12: 50177790-50177790 on Build GRCh38
rs777217378 Chr.12: 50177693-50177693 on Build GRCh38
rs755551433 Chr.12: 50177806-50177806 on Build GRCh38
rs766627352 Chr.12: 50177754-50177754 on Build GRCh38
rs138501981 Chr.12: 50177690-50177690 on Build GRCh38
rs957446269 Chr.12: 50177682-50177682 on Build GRCh38
rs201311819 Chr.12: 50177786-50177786 on Build GRCh38
rs766994071 Chr.12: 50177649-50177649 on Build GRCh38
rs761889209 Chr.12: 50177740-50177740 on Build GRCh38
rs755931465 Chr.12: 50177677-50177677 on Build GRCh38
rs755287834 Chr.12: 50177674-50177674 on Build GRCh38
rs779770726 Chr.12: 50177815-50177815 on Build GRCh38
rs776535993 Chr.12: 50177736-50177736 on Build GRCh38
rs769442487 Chr.12: 50177739-50177739 on Build GRCh38
rs756628778 Chr.12: 50177648-50177648 on Build GRCh38
rs750550657 Chr.12: 50177642-50177642 on Build GRCh38
rs913362354 Chr.12: 50177752-50177752 on Build GRCh38
rs770788879 Chr.12: 50177731-50177731 on Build GRCh38
rs746537443 Chr.12: 50177717-50177717 on Build GRCh38
rs139548333 Chr.12: 50177753-50177753 on Build GRCh38
rs760930378 Chr.12: 50177678-50177678 on Build GRCh38
rs529893508 Chr.12: 50177664-50177664 on Build GRCh38
rs745866453 Chr.12: 50177738-50177738 on Build GRCh38
rs779388299 Chr.12: 50177675-50177675 on Build GRCh38
rs765482409 Chr.12: 50177810-50177810 on Build GRCh38

View Full Product Details