Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74345105-74345330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP11A1
Entrez Gene ID
1583
Gene Name
cytochrome P450 family 11 subfamily A member 1
Gene Aliases
CYP11A, CYPXIA1, P450SCC
UniGene
Hs.303980
Gene Chromosome Location
Chr.15: 74337762-74367740 on Build GRCh38
Transcripts
NM_000781.2 NM_001099773.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5342623 c.526C>T p.R176W Chr.15: 74345143-74345143 on Build GRCh38
4675857 c.448G>T p.D150Y Chr.15: 74345221-74345221 on Build GRCh38
1301414 c.553C>T p.R185C Chr.15: 74345116-74345116 on Build GRCh38
5599368 c.529G>A p.D177N Chr.15: 74345140-74345140 on Build GRCh38
6355379 c.474G>C p.V158V Chr.15: 74345195-74345195 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762299364 Chr.15: 74345124-74345124 on Build GRCh38
rs773454116 Chr.15: 74345305-74345305 on Build GRCh38
rs149170405 Chr.15: 74345191-74345191 on Build GRCh38
rs200235389 Chr.15: 74345129-74345129 on Build GRCh38
rs774258847 Chr.15: 74345287-74345287 on Build GRCh38
rs769898237 Chr.15: 74345200-74345200 on Build GRCh38
rs374425401 Chr.15: 74345142-74345142 on Build GRCh38
rs117641050 Chr.15: 74345290-74345290 on Build GRCh38
rs757917103 Chr.15: 74345255-74345255 on Build GRCh38
rs766911998 Chr.15: 74345240-74345240 on Build GRCh38
rs757872565 Chr.15: 74345156-74345156 on Build GRCh38
rs752253559 Chr.15: 74345152-74345152 on Build GRCh38
rs751255184 Chr.15: 74345264-74345264 on Build GRCh38
rs370155933 Chr.15: 74345269-74345269 on Build GRCh38
rs775534320 Chr.15: 74345207-74345207 on Build GRCh38
rs749546182 Chr.15: 74345305-74345305 on Build GRCh38
rs745837781 Chr.15: 74345198-74345198 on Build GRCh38
rs150405116 Chr.15: 74345135-74345135 on Build GRCh38
rs772234288 Chr.15: 74345292-74345292 on Build GRCh38
rs777459993 Chr.15: 74345158-74345158 on Build GRCh38
rs58174009 Chr.15: 74345134-74345134 on Build GRCh38
rs749183607 Chr.15: 74345272-74345272 on Build GRCh38
rs890144526 Chr.15: 74345282-74345282 on Build GRCh38
rs763187434 Chr.15: 74345217-74345217 on Build GRCh38
rs138182745 Chr.15: 74345182-74345182 on Build GRCh38
rs774650073 Chr.15: 74345230-74345230 on Build GRCh38
rs549043326 Chr.15: 74345238-74345238 on Build GRCh38
rs202108906 Chr.15: 74345265-74345265 on Build GRCh38
rs767957985 Chr.15: 74345237-74345237 on Build GRCh38
rs571225194 Chr.15: 74345307-74345307 on Build GRCh38
rs879248075 Chr.15: 74345254-74345254 on Build GRCh38
rs769365856 Chr.15: 74345164-74345164 on Build GRCh38
rs778417752 Chr.15: 74345143-74345143 on Build GRCh38
rs768847979 Chr.15: 74345219-74345219 on Build GRCh38
rs756539849 Chr.15: 74345138-74345138 on Build GRCh38
rs559963981 Chr.15: 74345172-74345172 on Build GRCh38
rs763526130 Chr.15: 74345256-74345256 on Build GRCh38
rs745687786 Chr.15: 74345267-74345267 on Build GRCh38
rs746584608 Chr.15: 74345175-74345175 on Build GRCh38
rs141920628 Chr.15: 74345231-74345231 on Build GRCh38
rs531481036 Chr.15: 74345277-74345277 on Build GRCh38
rs754329273 Chr.15: 74345245-74345245 on Build GRCh38
rs780872909 Chr.15: 74345266-74345266 on Build GRCh38
rs1057073202 Chr.15: 74345308-74345308 on Build GRCh38

View Full Product Details