Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 11121635-11121902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCCS
Entrez Gene ID
3052
Gene Name
holocytochrome c synthase
Gene Aliases
CCHL, LSDMCA1, MCOPS7, MLS
UniGene
Hs.211571
Gene Chromosome Location
Chr.X: 11111286-11123086 on Build GRCh38
Transcripts
NM_001122608.2 NM_001171991.2 NM_005333.4

SNP Detail

SNP ID Chromosome Location
rs750492812 Chr.X: 11121804-11121804 on Build GRCh38
rs767566590 Chr.X: 11121786-11121786 on Build GRCh38
rs906220079 Chr.X: 11121864-11121864 on Build GRCh38
rs752236185 Chr.X: 11121825-11121825 on Build GRCh38
rs199575974 Chr.X: 11121700-11121700 on Build GRCh38
rs758677010 Chr.X: 11121806-11121806 on Build GRCh38
rs761396217 Chr.X: 11121708-11121708 on Build GRCh38
rs752336640 Chr.X: 11121841-11121842 on Build GRCh38
rs377026458 Chr.X: 11121836-11121836 on Build GRCh38
rs34228583 Chr.X: 11121762-11121762 on Build GRCh38
rs887015236 Chr.X: 11121790-11121790 on Build GRCh38
rs145499860 Chr.X: 11121766-11121766 on Build GRCh38
rs763383781 Chr.X: 11121827-11121828 on Build GRCh38
rs1027305580 Chr.X: 11121765-11121765 on Build GRCh38
rs189107814 Chr.X: 11121690-11121690 on Build GRCh38
rs764606786 Chr.X: 11121730-11121730 on Build GRCh38
rs139955716 Chr.X: 11121735-11121735 on Build GRCh38
rs748567469 Chr.X: 11121840-11121840 on Build GRCh38
rs372501217 Chr.X: 11121797-11121797 on Build GRCh38
rs181772486 Chr.X: 11121854-11121854 on Build GRCh38
rs765008200 Chr.X: 11121817-11121817 on Build GRCh38
rs747376781 Chr.X: 11121824-11121824 on Build GRCh38
rs1046078662 Chr.X: 11121838-11121838 on Build GRCh38
rs752520042 Chr.X: 11121787-11121787 on Build GRCh38
rs1001933308 Chr.X: 11121873-11121873 on Build GRCh38
rs751458949 Chr.X: 11121761-11121761 on Build GRCh38
rs757388811 Chr.X: 11121788-11121788 on Build GRCh38
rs775880525 Chr.X: 11121657-11121657 on Build GRCh38
rs201692478 Chr.X: 11121745-11121745 on Build GRCh38
rs762555651 Chr.X: 11121737-11121737 on Build GRCh38
rs776931746 Chr.X: 11121706-11121706 on Build GRCh38
rs1048305640 Chr.X: 11121741-11121741 on Build GRCh38

View Full Product Details