Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27355516-27355769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAP3K6
Entrez Gene ID
9064
Gene Name
mitogen-activated protein kinase kinase kinase 6
Gene Aliases
ASK2, MAPKKK6, MEKK6
UniGene
Hs.194694
Gene Chromosome Location
Chr.1: 27355179-27368037 on Build GRCh38
Transcripts
NM_001297609.1 NM_004672.4 XM_017002771.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
298470 c.3748A>T p.I1250F Chr.1: 27355685-27355685 on Build GRCh38
4700367 c.3758G>A p.R1253H Chr.1: 27355675-27355675 on Build GRCh38
6363260 c.3782G>T p.R1261L Chr.1: 27355675-27355675 on Build GRCh38
4872259 c.3754G>A p.A1252T Chr.1: 27355703-27355703 on Build GRCh38
5984902 c.3726C>T p.S1242S Chr.1: 27355731-27355731 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769707001 Chr.1: 27355685-27355685 on Build GRCh38
rs766148863 Chr.1: 27355706-27355706 on Build GRCh38
rs71638522 Chr.1: 27355722-27355722 on Build GRCh38
rs754225050 Chr.1: 27355735-27355735 on Build GRCh38
rs745966999 Chr.1: 27355675-27355675 on Build GRCh38
rs751036669 Chr.1: 27355707-27355707 on Build GRCh38
rs749448425 Chr.1: 27355657-27355657 on Build GRCh38
rs778916451 Chr.1: 27355664-27355664 on Build GRCh38
rs530633142 Chr.1: 27355548-27355548 on Build GRCh38
rs756425142 Chr.1: 27355641-27355641 on Build GRCh38
rs546863373 Chr.1: 27355684-27355684 on Build GRCh38
rs776410380 Chr.1: 27355678-27355678 on Build GRCh38
rs550648615 Chr.1: 27355600-27355600 on Build GRCh38
rs773144014 Chr.1: 27355688-27355688 on Build GRCh38
rs76570472 Chr.1: 27355710-27355710 on Build GRCh38
rs767117230 Chr.1: 27355718-27355718 on Build GRCh38
rs766950034 Chr.1: 27355665-27355665 on Build GRCh38
rs12742921 Chr.1: 27355635-27355635 on Build GRCh38
rs371911149 Chr.1: 27355652-27355652 on Build GRCh38
rs771156980 Chr.1: 27355663-27355663 on Build GRCh38
rs1020919674 Chr.1: 27355568-27355568 on Build GRCh38
rs768616209 Chr.1: 27355676-27355676 on Build GRCh38
rs778055953 Chr.1: 27355734-27355734 on Build GRCh38
rs757432362 Chr.1: 27355746-27355746 on Build GRCh38
rs774891853 Chr.1: 27355650-27355651 on Build GRCh38
rs375004232 Chr.1: 27355724-27355724 on Build GRCh38
rs1009485205 Chr.1: 27355567-27355575 on Build GRCh38
rs141031254 Chr.1: 27355726-27355726 on Build GRCh38
rs151081848 Chr.1: 27355608-27355608 on Build GRCh38
rs753301243 Chr.1: 27355638-27355638 on Build GRCh38
rs370697919 Chr.1: 27355695-27355695 on Build GRCh38
rs111630918 Chr.1: 27355676-27355676 on Build GRCh38
rs759709284 Chr.1: 27355648-27355648 on Build GRCh38

View Full Product Details