Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70958392-70958655 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1B1
Entrez Gene ID
525
Gene Name
ATPase H+ transporting V1 subunit B1
Gene Aliases
ATP6B1, RTA1B, VATB, VMA2, VPP3
UniGene
Hs.64173
Gene Chromosome Location
Chr.2: 70935868-70965431 on Build GRCh38
Transcripts
NM_001692.3 XM_011532907.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1022558 c.347C>T p.P116L Chr.2: 70958406-70958406 on Build GRCh38
5693602 c.363G>A p.M121I Chr.2: 70958422-70958422 on Build GRCh38
7109822 c.339A>G p.L113L Chr.2: 70958398-70958398 on Build GRCh38
4543670 c.341G>A p.R114Q Chr.2: 70958400-70958400 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781825701 Chr.2: 70958414-70958414 on Build GRCh38
rs781973207 Chr.2: 70958445-70958445 on Build GRCh38
rs201325403 Chr.2: 70958421-70958421 on Build GRCh38
rs374351693 Chr.2: 70958426-70958426 on Build GRCh38
rs996754913 Chr.2: 70958419-70958419 on Build GRCh38
rs782494901 Chr.2: 70958420-70958420 on Build GRCh38
rs538795304 Chr.2: 70958435-70958435 on Build GRCh38
rs1031704215 Chr.2: 70958515-70958515 on Build GRCh38
rs554966048 Chr.2: 70958568-70958568 on Build GRCh38
rs782025148 Chr.2: 70958450-70958450 on Build GRCh38
rs977829216 Chr.2: 70958512-70958512 on Build GRCh38
rs565946898 Chr.2: 70958472-70958472 on Build GRCh38
rs782247760 Chr.2: 70958448-70958448 on Build GRCh38
rs141698649 Chr.2: 70958547-70958547 on Build GRCh38
rs372947302 Chr.2: 70958452-70958452 on Build GRCh38
rs562711313 Chr.2: 70958468-70958468 on Build GRCh38
rs367614079 Chr.2: 70958449-70958449 on Build GRCh38
rs531801698 Chr.2: 70958489-70958489 on Build GRCh38
rs782372705 Chr.2: 70958466-70958466 on Build GRCh38
rs970213422 Chr.2: 70958614-70958614 on Build GRCh38
rs957682353 Chr.2: 70958565-70958565 on Build GRCh38
rs781865505 Chr.2: 70958475-70958475 on Build GRCh38
rs981635676 Chr.2: 70958619-70958619 on Build GRCh38
rs782564603 Chr.2: 70958429-70958429 on Build GRCh38
rs376250748 Chr.2: 70958456-70958456 on Build GRCh38
rs916064823 Chr.2: 70958606-70958606 on Build GRCh38
rs955337147 Chr.2: 70958441-70958441 on Build GRCh38
rs782086753 Chr.2: 70958462-70958462 on Build GRCh38
rs547564219 Chr.2: 70958443-70958443 on Build GRCh38

View Full Product Details