Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 769714-769987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLOD4
Entrez Gene ID
51031
Gene Name
glyoxalase domain containing 4
Gene Aliases
C17orf25, CGI-150, HC71
UniGene
Hs.279061
Gene Chromosome Location
Chr.17: 759309-782331 on Build GRCh38
Transcripts
NM_016080.3 XM_006721540.2 XM_017024711.1 XM_017024712.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4429178 c.829C>T p.R277* Chr.17: 769916-769916 on Build GRCh38
4980542 c.813C>T p.V271V Chr.17: 769932-769932 on Build GRCh38
4429177 c.784C>T p.R262* Chr.17: 769916-769916 on Build GRCh38
3787413 c.750A>T p.G250G Chr.17: 769950-769950 on Build GRCh38
6671679 c.861C>A p.S287R Chr.17: 769884-769884 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs17851349 Chr.17: 769899-769899 on Build GRCh38
rs999212353 Chr.17: 769906-769906 on Build GRCh38
rs769479560 Chr.17: 769909-769909 on Build GRCh38
rs11663 Chr.17: 769950-769950 on Build GRCh38
rs766236555 Chr.17: 769829-769829 on Build GRCh38
rs184466050 Chr.17: 769916-769916 on Build GRCh38
rs143261573 Chr.17: 769843-769843 on Build GRCh38
rs139221941 Chr.17: 769748-769748 on Build GRCh38
rs767711047 Chr.17: 769938-769938 on Build GRCh38
rs753988099 Chr.17: 769822-769822 on Build GRCh38
rs778011488 Chr.17: 769807-769807 on Build GRCh38
rs544906354 Chr.17: 769743-769743 on Build GRCh38
rs376309803 Chr.17: 769841-769841 on Build GRCh38
rs542245318 Chr.17: 769892-769892 on Build GRCh38
rs960455532 Chr.17: 769907-769907 on Build GRCh38
rs761994898 Chr.17: 769864-769865 on Build GRCh38
rs114814106 Chr.17: 769915-769915 on Build GRCh38
rs749838246 Chr.17: 769886-769886 on Build GRCh38
rs751683450 Chr.17: 769832-769832 on Build GRCh38
rs756241718 Chr.17: 769880-769880 on Build GRCh38
rs373166347 Chr.17: 769804-769805 on Build GRCh38
rs139770786 Chr.17: 769953-769953 on Build GRCh38
rs1046655436 Chr.17: 769835-769835 on Build GRCh38
rs115211226 Chr.17: 769928-769928 on Build GRCh38
rs754254427 Chr.17: 769955-769955 on Build GRCh38
rs776496105 Chr.17: 769902-769902 on Build GRCh38
rs889678468 Chr.17: 769814-769814 on Build GRCh38
rs778227509 Chr.17: 769883-769883 on Build GRCh38
rs773342760 Chr.17: 769820-769820 on Build GRCh38
rs762634820 Chr.17: 769821-769821 on Build GRCh38
rs746588800 Chr.17: 769898-769898 on Build GRCh38
rs923250319 Chr.17: 769752-769752 on Build GRCh38
rs148206481 Chr.17: 769920-769920 on Build GRCh38
rs141571460 Chr.17: 769932-769932 on Build GRCh38
rs752768207 Chr.17: 769869-769869 on Build GRCh38
rs757518800 Chr.17: 769890-769890 on Build GRCh38
rs377262154 Chr.17: 769751-769751 on Build GRCh38

View Full Product Details