Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38088697-38088932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAIAP2L2
Entrez Gene ID
80115
Gene Name
BAI1 associated protein 2 like 2
Gene Aliases
-
UniGene
Hs.474822
Gene Chromosome Location
Chr.22: 38084889-38110944 on Build GRCh38
Transcripts
NM_025045.5 XM_005261751.4 XM_011530379.2 XM_011530380.2 XM_011530381.2 XM_011530382.2 XM_011530383.2 XM_011530384.2 XM_011530386.2 XM_011530387.2 XM_011530388.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6609802 c.1015G>A p.A339T Chr.22: 38088851-38088851 on Build GRCh38
4387747 c.1083C>T p.N361N Chr.22: 38088783-38088783 on Build GRCh38
4104051 c.1014C>T p.G338G Chr.22: 38088852-38088852 on Build GRCh38
5636591 c.1050C>T p.D350D Chr.22: 38088816-38088816 on Build GRCh38
4104052 c.960G>A p.P320P Chr.22: 38088906-38088906 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs994505929 Chr.22: 38088915-38088915 on Build GRCh38
rs767099374 Chr.22: 38088893-38088893 on Build GRCh38
rs947918822 Chr.22: 38088787-38088787 on Build GRCh38
rs987623777 Chr.22: 38088739-38088739 on Build GRCh38
rs764049790 Chr.22: 38088910-38088910 on Build GRCh38
rs772656015 Chr.22: 38088781-38088781 on Build GRCh38
rs780360460 Chr.22: 38088858-38088858 on Build GRCh38
rs747393218 Chr.22: 38088859-38088859 on Build GRCh38
rs753904518 Chr.22: 38088824-38088824 on Build GRCh38
rs759195108 Chr.22: 38088890-38088890 on Build GRCh38
rs376964284 Chr.22: 38088753-38088753 on Build GRCh38
rs771221773 Chr.22: 38088754-38088754 on Build GRCh38
rs762461084 Chr.22: 38088783-38088783 on Build GRCh38
rs373008506 Chr.22: 38088751-38088751 on Build GRCh38
rs201879183 Chr.22: 38088748-38088748 on Build GRCh38
rs759166798 Chr.22: 38088807-38088807 on Build GRCh38
rs778920958 Chr.22: 38088832-38088832 on Build GRCh38
rs757355982 Chr.22: 38088825-38088825 on Build GRCh38
rs746082756 Chr.22: 38088837-38088837 on Build GRCh38
rs775367064 Chr.22: 38088894-38088894 on Build GRCh38
rs779202347 Chr.22: 38088757-38088757 on Build GRCh38
rs747232727 Chr.22: 38088906-38088913 on Build GRCh38
rs773780836 Chr.22: 38088878-38088878 on Build GRCh38
rs895622217 Chr.22: 38088800-38088800 on Build GRCh38
rs776063111 Chr.22: 38088721-38088721 on Build GRCh38
rs939222100 Chr.22: 38088782-38088782 on Build GRCh38
rs769089800 Chr.22: 38088863-38088863 on Build GRCh38
rs916395818 Chr.22: 38088784-38088784 on Build GRCh38
rs772393636 Chr.22: 38088720-38088720 on Build GRCh38
rs937200968 Chr.22: 38088773-38088773 on Build GRCh38
rs765877682 Chr.22: 38088792-38088792 on Build GRCh38
rs758635517 Chr.22: 38088762-38088763 on Build GRCh38
rs775771080 Chr.22: 38088770-38088770 on Build GRCh38
rs778193520 Chr.22: 38088848-38088886 on Build GRCh38
rs201140269 Chr.22: 38088861-38088861 on Build GRCh38
rs372692732 Chr.22: 38088876-38088876 on Build GRCh38
rs751218638 Chr.22: 38088801-38088801 on Build GRCh38
rs769177938 Chr.22: 38088780-38088780 on Build GRCh38
rs369670314 Chr.22: 38088723-38088723 on Build GRCh38
rs748554012 Chr.22: 38088756-38088757 on Build GRCh38
rs374885175 Chr.22: 38088746-38088746 on Build GRCh38
rs764709043 Chr.22: 38088722-38088722 on Build GRCh38
rs780915187 Chr.22: 38088747-38088747 on Build GRCh38
rs746146034 Chr.22: 38088764-38088764 on Build GRCh38
rs755505438 Chr.22: 38088717-38088718 on Build GRCh38
rs1048793535 Chr.22: 38088821-38088821 on Build GRCh38
rs371552367 Chr.22: 38088743-38088743 on Build GRCh38
rs927201443 Chr.22: 38088761-38088761 on Build GRCh38
rs760552564 Chr.22: 38088903-38088903 on Build GRCh38
rs767321736 Chr.22: 38088810-38088810 on Build GRCh38
rs370080453 Chr.22: 38088815-38088815 on Build GRCh38
rs764141561 Chr.22: 38088823-38088823 on Build GRCh38
rs528397864 Chr.22: 38088771-38088771 on Build GRCh38
rs779356293 Chr.22: 38088719-38088719 on Build GRCh38
rs753816499 Chr.22: 38088913-38088913 on Build GRCh38
rs756067567 Chr.22: 38088819-38088819 on Build GRCh38
rs748718248 Chr.22: 38088877-38088877 on Build GRCh38
rs758592823 Chr.22: 38088849-38088849 on Build GRCh38
rs772473669 Chr.22: 38088765-38088765 on Build GRCh38
rs762549099 Chr.22: 38088735-38088735 on Build GRCh38

View Full Product Details