Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 71538186-71538678 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRTAP5-8
Entrez Gene ID
57830
Gene Name
keratin associated protein 5-8
Gene Aliases
KRTAP5-2, KRTAP5.8, UHSKerB
UniGene
Hs.445245
Gene Chromosome Location
Chr.11: 71538025-71539207 on Build GRCh38
Transcripts
NM_021046.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5956778 c.333A>G p.K111K Chr.11: 71538388-71538388 on Build GRCh38
2044606 c.452C>T p.P151L Chr.11: 71538507-71538507 on Build GRCh38
931590 c.457T>A p.C153S Chr.11: 71538512-71538512 on Build GRCh38
3452930 c.245C>T p.S82F Chr.11: 71538300-71538300 on Build GRCh38
6232692 c.429T>C p.C143C Chr.11: 71538484-71538484 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534497246 Chr.11: 71538289-71538289 on Build GRCh38
rs1018657487 Chr.11: 71538280-71538280 on Build GRCh38
rs868514964 Chr.11: 71538550-71538550 on Build GRCh38
rs190660192 Chr.11: 71538233-71538233 on Build GRCh38
rs200888449 Chr.11: 71538604-71538604 on Build GRCh38
rs769033508 Chr.11: 71538570-71538581 on Build GRCh38
rs748767379 Chr.11: 71538573-71538574 on Build GRCh38
rs7101792 Chr.11: 71538340-71538340 on Build GRCh38
rs779049580 Chr.11: 71538230-71538230 on Build GRCh38
rs746581843 Chr.11: 71538597-71538597 on Build GRCh38
rs751475151 Chr.11: 71538266-71538266 on Build GRCh38
rs377076492 Chr.11: 71538530-71538530 on Build GRCh38
rs879109427 Chr.11: 71538388-71538388 on Build GRCh38
rs765031409 Chr.11: 71538615-71538615 on Build GRCh38
rs747469393 Chr.11: 71538300-71538300 on Build GRCh38
rs781590727 Chr.11: 71538310-71538310 on Build GRCh38
rs758735417 Chr.11: 71538446-71538447 on Build GRCh38
rs878874424 Chr.11: 71538433-71538433 on Build GRCh38
rs771572907 Chr.11: 71538588-71538588 on Build GRCh38
rs767454491 Chr.11: 71538364-71538364 on Build GRCh38
rs548540927 Chr.11: 71538276-71538276 on Build GRCh38
rs879079541 Chr.11: 71538484-71538484 on Build GRCh38
rs747522411 Chr.11: 71538238-71538238 on Build GRCh38
rs183256246 Chr.11: 71538247-71538247 on Build GRCh38
rs377029760 Chr.11: 71538326-71538326 on Build GRCh38
rs572291847 Chr.11: 71538242-71538242 on Build GRCh38
rs541819431 Chr.11: 71538232-71538232 on Build GRCh38
rs752932165 Chr.11: 71538279-71538279 on Build GRCh38
rs772622043 Chr.11: 71538596-71538596 on Build GRCh38
rs144266749 Chr.11: 71538419-71538419 on Build GRCh38
rs374587723 Chr.11: 71538512-71538512 on Build GRCh38
rs200517331 Chr.11: 71538460-71538460 on Build GRCh38
rs11234083 Chr.11: 71538399-71538399 on Build GRCh38
rs746376460 Chr.11: 71538446-71538446 on Build GRCh38
rs777866571 Chr.11: 71538297-71538297 on Build GRCh38
rs760565042 Chr.11: 71538496-71538496 on Build GRCh38
rs759868215 Chr.11: 71538248-71538248 on Build GRCh38
rs372455775 Chr.11: 71538412-71538412 on Build GRCh38
rs878929953 Chr.11: 71538520-71538520 on Build GRCh38
rs781701703 Chr.11: 71538570-71538571 on Build GRCh38
rs11234084 Chr.11: 71538486-71538486 on Build GRCh38
rs773138709 Chr.11: 71538492-71538492 on Build GRCh38
rs751787965 Chr.11: 71538559-71538559 on Build GRCh38
rs898993984 Chr.11: 71538218-71538218 on Build GRCh38
rs779234678 Chr.11: 71538234-71538263 on Build GRCh38
rs376137869 Chr.11: 71538586-71538586 on Build GRCh38
rs375732177 Chr.11: 71538211-71538211 on Build GRCh38
rs774020725 Chr.11: 71538343-71538343 on Build GRCh38
rs532438179 Chr.11: 71538457-71538486 on Build GRCh38
rs940570428 Chr.11: 71538574-71538574 on Build GRCh38
rs756432687 Chr.11: 71538452-71538452 on Build GRCh38
rs756710318 Chr.11: 71538573-71538573 on Build GRCh38
rs774568125 Chr.11: 71538617-71538617 on Build GRCh38
rs775350808 Chr.11: 71538252-71538252 on Build GRCh38
rs766169799 Chr.11: 71538375-71538375 on Build GRCh38
rs750628947 Chr.11: 71538262-71538291 on Build GRCh38
rs762540041 Chr.11: 71538253-71538253 on Build GRCh38
rs761839979 Chr.11: 71538267-71538267 on Build GRCh38
rs950203475 Chr.11: 71538450-71538450 on Build GRCh38
rs758646337 Chr.11: 71538396-71538396 on Build GRCh38
rs760502824 Chr.11: 71538369-71538369 on Build GRCh38
rs750901242 Chr.11: 71538571-71538571 on Build GRCh38
rs747476855 Chr.11: 71538565-71538567 on Build GRCh38
rs764296730 Chr.11: 71538549-71538549 on Build GRCh38
rs201312529 Chr.11: 71538501-71538501 on Build GRCh38
rs199734215 Chr.11: 71538564-71538564 on Build GRCh38
rs776393399 Chr.11: 71538534-71538534 on Build GRCh38
rs574564297 Chr.11: 71538476-71538476 on Build GRCh38
rs748901042 Chr.11: 71538465-71538465 on Build GRCh38
rs778022898 Chr.11: 71538400-71538400 on Build GRCh38
rs369043826 Chr.11: 71538484-71538486 on Build GRCh38
rs564299923 Chr.11: 71538249-71538249 on Build GRCh38
rs373878010 Chr.11: 71538646-71538646 on Build GRCh38
rs370033683 Chr.11: 71538540-71538540 on Build GRCh38
rs114280911 Chr.11: 71538656-71538656 on Build GRCh38
rs531354782 Chr.11: 71538257-71538257 on Build GRCh38
rs781044889 Chr.11: 71538207-71538207 on Build GRCh38
rs770115058 Chr.11: 71538316-71538316 on Build GRCh38
rs11234082 Chr.11: 71538312-71538312 on Build GRCh38
rs771447989 Chr.11: 71538306-71538306 on Build GRCh38
rs376584900 Chr.11: 71538345-71538345 on Build GRCh38
rs200162819 Chr.11: 71538483-71538483 on Build GRCh38
rs112605265 Chr.11: 71538425-71538425 on Build GRCh38
rs574501468 Chr.11: 71538231-71538231 on Build GRCh38
rs757305365 Chr.11: 71538567-71538567 on Build GRCh38
rs562607417 Chr.11: 71538261-71538261 on Build GRCh38
rs369249854 Chr.11: 71538336-71538336 on Build GRCh38
rs767497901 Chr.11: 71538570-71538570 on Build GRCh38
rs776617609 Chr.11: 71538609-71538609 on Build GRCh38
rs755247699 Chr.11: 71538213-71538213 on Build GRCh38
rs774184326 Chr.11: 71538475-71538475 on Build GRCh38
rs575845765 Chr.11: 71538618-71538618 on Build GRCh38
rs376694979 Chr.11: 71538472-71538472 on Build GRCh38
rs755457639 Chr.11: 71538585-71538585 on Build GRCh38
rs538408077 Chr.11: 71538385-71538385 on Build GRCh38
rs867693463 Chr.11: 71538283-71538283 on Build GRCh38
rs755898933 Chr.11: 71538277-71538277 on Build GRCh38
rs749634984 Chr.11: 71538578-71538578 on Build GRCh38
rs759314691 Chr.11: 71538613-71538613 on Build GRCh38
rs540274094 Chr.11: 71538471-71538471 on Build GRCh38
rs550667548 Chr.11: 71538259-71538259 on Build GRCh38
rs757348384 Chr.11: 71538414-71538414 on Build GRCh38
rs865962074 Chr.11: 71538575-71538575 on Build GRCh38
rs755215255 Chr.11: 71538217-71538217 on Build GRCh38
rs530057198 Chr.11: 71538263-71538263 on Build GRCh38
rs546079349 Chr.11: 71538576-71538576 on Build GRCh38
rs1029171826 Chr.11: 71538237-71538237 on Build GRCh38
rs750447332 Chr.11: 71538653-71538653 on Build GRCh38
rs567085723 Chr.11: 71538288-71538288 on Build GRCh38
rs751607151 Chr.11: 71538404-71538404 on Build GRCh38

View Full Product Details