Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 73297149-73297649 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
P2RY6
Entrez Gene ID
5031
Gene Name
pyrimidinergic receptor P2Y6
Gene Aliases
P2Y6
UniGene
Hs.16362
Gene Chromosome Location
Chr.11: 73264503-73298625 on Build GRCh38
Transcripts
NM_001277204.1 NM_001277205.1 NM_001277206.1 NM_001277207.1 NM_001277208.1 NM_176796.2 NM_176797.2 NM_176798.2 XM_005274022.3 XM_006718571.3 XM_011545076.2 XM_011545077.2 XM_011545079.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4900153 c.720G>A p.M240I Chr.11: 73297238-73297238 on Build GRCh38
3769439 c.779C>A p.T260K Chr.11: 73297297-73297297 on Build GRCh38
6736522 c.860G>A p.R287Q Chr.11: 73297378-73297378 on Build GRCh38
3687632 c.723C>A p.A241A Chr.11: 73297241-73297241 on Build GRCh38
6736518 c.672C>T p.G224G Chr.11: 73297190-73297190 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766899289 Chr.11: 73297343-73297343 on Build GRCh38
rs781206538 Chr.11: 73297258-73297258 on Build GRCh38
rs913720145 Chr.11: 73297192-73297192 on Build GRCh38
rs143057636 Chr.11: 73297211-73297211 on Build GRCh38
rs761394326 Chr.11: 73297337-73297337 on Build GRCh38
rs763374936 Chr.11: 73297326-73297326 on Build GRCh38
rs889880239 Chr.11: 73297529-73297529 on Build GRCh38
rs139336490 Chr.11: 73297446-73297446 on Build GRCh38
rs755479716 Chr.11: 73297531-73297531 on Build GRCh38
rs368917743 Chr.11: 73297401-73297401 on Build GRCh38
rs988017987 Chr.11: 73297408-73297408 on Build GRCh38
rs200512829 Chr.11: 73297329-73297329 on Build GRCh38
rs377684373 Chr.11: 73297457-73297457 on Build GRCh38
rs766361758 Chr.11: 73297430-73297430 on Build GRCh38
rs138593381 Chr.11: 73297216-73297216 on Build GRCh38
rs765808774 Chr.11: 73297495-73297495 on Build GRCh38
rs539417498 Chr.11: 73297195-73297195 on Build GRCh38
rs911799901 Chr.11: 73297178-73297178 on Build GRCh38
rs138018794 Chr.11: 73297187-73297187 on Build GRCh38
rs753700391 Chr.11: 73297347-73297347 on Build GRCh38
rs554148229 Chr.11: 73297452-73297452 on Build GRCh38
rs763902567 Chr.11: 73297328-73297328 on Build GRCh38
rs770561514 Chr.11: 73297204-73297204 on Build GRCh38
rs756712823 Chr.11: 73297254-73297254 on Build GRCh38
rs566488664 Chr.11: 73297179-73297179 on Build GRCh38
rs754754550 Chr.11: 73297357-73297357 on Build GRCh38
rs144114236 Chr.11: 73297506-73297506 on Build GRCh38
rs1049897154 Chr.11: 73297520-73297520 on Build GRCh38
rs748518697 Chr.11: 73297544-73297544 on Build GRCh38
rs552250343 Chr.11: 73297546-73297546 on Build GRCh38
rs776579171 Chr.11: 73297496-73297496 on Build GRCh38
rs201782854 Chr.11: 73297278-73297278 on Build GRCh38
rs546534644 Chr.11: 73297171-73297171 on Build GRCh38
rs747171091 Chr.11: 73297314-73297314 on Build GRCh38
rs61741436 Chr.11: 73297344-73297344 on Build GRCh38
rs150908498 Chr.11: 73297444-73297444 on Build GRCh38
rs772838232 Chr.11: 73297461-73297461 on Build GRCh38
rs765196072 Chr.11: 73297498-73297498 on Build GRCh38
rs755937132 Chr.11: 73297265-73297265 on Build GRCh38
rs558670933 Chr.11: 73297193-73297193 on Build GRCh38
rs779936701 Chr.11: 73297346-73297346 on Build GRCh38
rs576174370 Chr.11: 73297377-73297377 on Build GRCh38
rs772321901 Chr.11: 73297287-73297287 on Build GRCh38
rs373469202 Chr.11: 73297441-73297441 on Build GRCh38
rs758071334 Chr.11: 73297242-73297242 on Build GRCh38
rs780236031 Chr.11: 73297526-73297526 on Build GRCh38
rs371644059 Chr.11: 73297361-73297361 on Build GRCh38
rs576807939 Chr.11: 73297345-73297345 on Build GRCh38
rs200821631 Chr.11: 73297450-73297450 on Build GRCh38
rs374839880 Chr.11: 73297396-73297396 on Build GRCh38
rs113910844 Chr.11: 73297567-73297567 on Build GRCh38
rs749360133 Chr.11: 73297530-73297530 on Build GRCh38
rs763812571 Chr.11: 73297249-73297249 on Build GRCh38
rs756297541 Chr.11: 73297514-73297514 on Build GRCh38
rs60848410 Chr.11: 73297304-73297304 on Build GRCh38
rs142217062 Chr.11: 73297443-73297443 on Build GRCh38
rs141420011 Chr.11: 73297170-73297170 on Build GRCh38
rs777198362 Chr.11: 73297318-73297318 on Build GRCh38
rs149614765 Chr.11: 73297451-73297451 on Build GRCh38
rs758009142 Chr.11: 73297180-73297198 on Build GRCh38
rs986782798 Chr.11: 73297175-73297175 on Build GRCh38
rs760640867 Chr.11: 73297218-73297218 on Build GRCh38
rs746765540 Chr.11: 73297432-73297434 on Build GRCh38
rs771071916 Chr.11: 73297363-73297363 on Build GRCh38
rs141681227 Chr.11: 73297228-73297228 on Build GRCh38
rs193163379 Chr.11: 73297501-73297501 on Build GRCh38
rs751116416 Chr.11: 73297250-73297250 on Build GRCh38
rs778069030 Chr.11: 73297360-73297360 on Build GRCh38
rs150383317 Chr.11: 73297180-73297180 on Build GRCh38
rs149089615 Chr.11: 73297208-73297208 on Build GRCh38
rs754771310 Chr.11: 73297433-73297433 on Build GRCh38
rs769079087 Chr.11: 73297382-73297382 on Build GRCh38
rs940241881 Chr.11: 73297311-73297311 on Build GRCh38
rs779471492 Chr.11: 73297541-73297541 on Build GRCh38
rs542361416 Chr.11: 73297400-73297400 on Build GRCh38
rs139384226 Chr.11: 73297321-73297321 on Build GRCh38
rs115479773 Chr.11: 73297315-73297315 on Build GRCh38
rs760191570 Chr.11: 73297462-73297462 on Build GRCh38
rs745728953 Chr.11: 73297262-73297262 on Build GRCh38
rs373254348 Chr.11: 73297202-73297202 on Build GRCh38
rs569014304 Chr.11: 73297234-73297234 on Build GRCh38
rs753716744 Chr.11: 73297432-73297432 on Build GRCh38
rs1020831170 Chr.11: 73297623-73297623 on Build GRCh38
rs780956300 Chr.11: 73297190-73297190 on Build GRCh38
rs745917913 Chr.11: 73297375-73297375 on Build GRCh38
rs375926613 Chr.11: 73297233-73297233 on Build GRCh38
rs780714539 Chr.11: 73297449-73297449 on Build GRCh38
rs760173562 Chr.11: 73297412-73297412 on Build GRCh38
rs537555142 Chr.11: 73297583-73297583 on Build GRCh38
rs763428362 Chr.11: 73297378-73297378 on Build GRCh38
rs779580266 Chr.11: 73297331-73297331 on Build GRCh38
rs766976278 Chr.11: 73297219-73297219 on Build GRCh38
rs781080996 Chr.11: 73297371-73297371 on Build GRCh38
rs564797664 Chr.11: 73297497-73297497 on Build GRCh38
rs769977823 Chr.11: 73297447-73297447 on Build GRCh38
rs772854464 Chr.11: 73297549-73297549 on Build GRCh38
rs773575254 Chr.11: 73297500-73297500 on Build GRCh38
rs368281933 Chr.11: 73297317-73297317 on Build GRCh38
rs779777489 Chr.11: 73297276-73297276 on Build GRCh38
rs562631177 Chr.11: 73297376-73297376 on Build GRCh38
rs369188404 Chr.11: 73297319-73297319 on Build GRCh38
rs372078262 Chr.11: 73297325-73297325 on Build GRCh38
rs1022803687 Chr.11: 73297283-73297283 on Build GRCh38
rs750610106 Chr.11: 73297508-73297508 on Build GRCh38
rs879567549 Chr.11: 73297612-73297612 on Build GRCh38
rs746623384 Chr.11: 73297557-73297557 on Build GRCh38

View Full Product Details