Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 78436118-78436638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NARS2
Entrez Gene ID
79731
Gene Name
asparaginyl-tRNA synthetase 2, mitochondrial (putative)
Gene Aliases
DFNB94, SLM5, asnRS
UniGene
Hs.503389
Gene Chromosome Location
Chr.11: 78435961-78574864 on Build GRCh38
Transcripts
NM_001243251.1 NM_024678.5 XM_011545253.1 XM_017018303.1 XM_017018304.1

SNP Detail

SNP ID Chromosome Location
rs1003654865 Chr.11: 78436218-78436218 on Build GRCh38
rs939411680 Chr.11: 78436574-78436574 on Build GRCh38
rs1028076459 Chr.11: 78436451-78436451 on Build GRCh38
rs574998770 Chr.11: 78436415-78436415 on Build GRCh38
rs73504911 Chr.11: 78436471-78436471 on Build GRCh38
rs928028101 Chr.11: 78436567-78436567 on Build GRCh38
rs531692627 Chr.11: 78436572-78436572 on Build GRCh38
rs547041422 Chr.11: 78436181-78436181 on Build GRCh38
rs913956821 Chr.11: 78436410-78436410 on Build GRCh38
rs113652771 Chr.11: 78436229-78436229 on Build GRCh38
rs568169874 Chr.11: 78436339-78436339 on Build GRCh38
rs771436826 Chr.11: 78436487-78436487 on Build GRCh38
rs184118063 Chr.11: 78436470-78436470 on Build GRCh38
rs527590896 Chr.11: 78436154-78436154 on Build GRCh38
rs939983144 Chr.11: 78436486-78436486 on Build GRCh38
rs770108388 Chr.11: 78436375-78436375 on Build GRCh38
rs879424598 Chr.11: 78436184-78436187 on Build GRCh38
rs146091248 Chr.11: 78436394-78436394 on Build GRCh38
rs7934948 Chr.11: 78436328-78436328 on Build GRCh38
rs1005742699 Chr.11: 78436264-78436264 on Build GRCh38
rs901349366 Chr.11: 78436458-78436458 on Build GRCh38
rs538009675 Chr.11: 78436389-78436389 on Build GRCh38
rs940032841 Chr.11: 78436180-78436180 on Build GRCh38
rs1022972232 Chr.11: 78436343-78436343 on Build GRCh38
rs142298861 Chr.11: 78436347-78436347 on Build GRCh38
rs190672535 Chr.11: 78436527-78436527 on Build GRCh38
rs895277991 Chr.11: 78436604-78436604 on Build GRCh38
rs375230339 Chr.11: 78436151-78436151 on Build GRCh38
rs180985602 Chr.11: 78436424-78436424 on Build GRCh38
rs541616138 Chr.11: 78436597-78436597 on Build GRCh38
rs973128003 Chr.11: 78436541-78436541 on Build GRCh38
rs909842296 Chr.11: 78436171-78436171 on Build GRCh38
rs1005195292 Chr.11: 78436499-78436499 on Build GRCh38
rs970001674 Chr.11: 78436334-78436334 on Build GRCh38
rs547643998 Chr.11: 78436266-78436266 on Build GRCh38
rs151249725 Chr.11: 78436529-78436530 on Build GRCh38
rs1037088980 Chr.11: 78436213-78436213 on Build GRCh38
rs569780008 Chr.11: 78436610-78436610 on Build GRCh38
rs936665750 Chr.11: 78436353-78436353 on Build GRCh38
rs976505095 Chr.11: 78436440-78436440 on Build GRCh38
rs554075537 Chr.11: 78436453-78436453 on Build GRCh38
rs570483528 Chr.11: 78436288-78436288 on Build GRCh38
rs992632441 Chr.11: 78436305-78436305 on Build GRCh38
rs953880936 Chr.11: 78436463-78436463 on Build GRCh38
rs996469147 Chr.11: 78436190-78436190 on Build GRCh38
rs138637786 Chr.11: 78436477-78436477 on Build GRCh38
rs906599560 Chr.11: 78436215-78436215 on Build GRCh38
rs1028443803 Chr.11: 78436205-78436205 on Build GRCh38
rs1055223605 Chr.11: 78436599-78436599 on Build GRCh38
rs961521912 Chr.11: 78436303-78436303 on Build GRCh38

View Full Product Details