Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 86264060-86264573 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EED
Entrez Gene ID
8726
Gene Name
embryonic ectoderm development
Gene Aliases
HEED, WAIT1
UniGene
Hs.503510
Gene Chromosome Location
Chr.11: 86244384-86285420 on Build GRCh38
Transcripts
NM_001308007.1 NM_003797.4 XM_005274373.2 XM_011545330.1 XM_011545331.1 XM_017018512.1 XM_017018513.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7348966 c.646C>T p.R216* Chr.11: 86264183-86264183 on Build GRCh38
7339652 c.688G>A p.G230R Chr.11: 86264225-86264225 on Build GRCh38
6935404 c.647G>A p.R216Q Chr.11: 86264184-86264184 on Build GRCh38
6254949 c.647G>T p.R216L Chr.11: 86264184-86264184 on Build GRCh38
6651703 c.692G>A p.G231D Chr.11: 86264229-86264229 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1046315479 Chr.11: 86264500-86264500 on Build GRCh38
rs975478034 Chr.11: 86264343-86264343 on Build GRCh38
rs1007811142 Chr.11: 86264485-86264485 on Build GRCh38
rs1022563224 Chr.11: 86264430-86264430 on Build GRCh38
rs796792497 Chr.11: 86264454-86264455 on Build GRCh38
rs966224755 Chr.11: 86264119-86264119 on Build GRCh38
rs750427382 Chr.11: 86264470-86264470 on Build GRCh38
rs146233392 Chr.11: 86264334-86264334 on Build GRCh38
rs892648669 Chr.11: 86264348-86264348 on Build GRCh38
rs756861839 Chr.11: 86264135-86264135 on Build GRCh38
rs906316193 Chr.11: 86264507-86264507 on Build GRCh38
rs948825972 Chr.11: 86264487-86264487 on Build GRCh38
rs748705884 Chr.11: 86264158-86264158 on Build GRCh38
rs372966988 Chr.11: 86264263-86264263 on Build GRCh38
rs975445564 Chr.11: 86264100-86264100 on Build GRCh38
rs764664651 Chr.11: 86264135-86264138 on Build GRCh38
rs745402003 Chr.11: 86264148-86264148 on Build GRCh38
rs763050365 Chr.11: 86264282-86264282 on Build GRCh38
rs769108276 Chr.11: 86264156-86264156 on Build GRCh38
rs180973790 Chr.11: 86264118-86264118 on Build GRCh38
rs763049552 Chr.11: 86264362-86264362 on Build GRCh38
rs759722952 Chr.11: 86264231-86264231 on Build GRCh38
rs972246822 Chr.11: 86264547-86264547 on Build GRCh38
rs74893075 Chr.11: 86264127-86264127 on Build GRCh38
rs941620186 Chr.11: 86264427-86264427 on Build GRCh38
rs78475799 Chr.11: 86264295-86264295 on Build GRCh38
rs35685239 Chr.11: 86264439-86264440 on Build GRCh38
rs906527117 Chr.11: 86264324-86264324 on Build GRCh38
rs1022103204 Chr.11: 86264333-86264333 on Build GRCh38
rs971830502 Chr.11: 86264125-86264125 on Build GRCh38
rs574255375 Chr.11: 86264389-86264389 on Build GRCh38
rs761139323 Chr.11: 86264135-86264137 on Build GRCh38
rs578082247 Chr.11: 86264146-86264146 on Build GRCh38
rs772475657 Chr.11: 86264159-86264159 on Build GRCh38
rs774811170 Chr.11: 86264157-86264157 on Build GRCh38
rs147153744 Chr.11: 86264188-86264188 on Build GRCh38
rs1052513757 Chr.11: 86264338-86264338 on Build GRCh38
rs752158739 Chr.11: 86264491-86264491 on Build GRCh38
rs751321344 Chr.11: 86264286-86264286 on Build GRCh38
rs71468992 Chr.11: 86264453-86264454 on Build GRCh38
rs776804989 Chr.11: 86264230-86264230 on Build GRCh38
rs147424448 Chr.11: 86264446-86264447 on Build GRCh38
rs199949721 Chr.11: 86264457-86264457 on Build GRCh38
rs1011051767 Chr.11: 86264421-86264421 on Build GRCh38
rs140126645 Chr.11: 86264308-86264309 on Build GRCh38
rs780735694 Chr.11: 86264303-86264303 on Build GRCh38
rs908526979 Chr.11: 86264407-86264407 on Build GRCh38
rs201509894 Chr.11: 86264439-86264439 on Build GRCh38
rs922673055 Chr.11: 86264388-86264388 on Build GRCh38
rs982839955 Chr.11: 86264401-86264401 on Build GRCh38
rs763881681 Chr.11: 86264283-86264283 on Build GRCh38
rs756012975 Chr.11: 86264476-86264476 on Build GRCh38
rs758244598 Chr.11: 86264112-86264112 on Build GRCh38
rs199568747 Chr.11: 86264440-86264441 on Build GRCh38
rs749952849 Chr.11: 86264269-86264269 on Build GRCh38
rs965725614 Chr.11: 86264498-86264498 on Build GRCh38
rs933973783 Chr.11: 86264107-86264107 on Build GRCh38
rs1045085324 Chr.11: 86264320-86264320 on Build GRCh38
rs1044802251 Chr.11: 86264266-86264266 on Build GRCh38
rs779871439 Chr.11: 86264496-86264496 on Build GRCh38
rs919112787 Chr.11: 86264202-86264202 on Build GRCh38
rs537879806 Chr.11: 86264453-86264453 on Build GRCh38
rs370092867 Chr.11: 86264145-86264145 on Build GRCh38
rs766523985 Chr.11: 86264203-86264203 on Build GRCh38
rs143650909 Chr.11: 86264186-86264186 on Build GRCh38
rs538340633 Chr.11: 86264092-86264092 on Build GRCh38
rs572894892 Chr.11: 86264315-86264315 on Build GRCh38
rs955642140 Chr.11: 86264393-86264393 on Build GRCh38
rs922608301 Chr.11: 86264105-86264105 on Build GRCh38
rs964119953 Chr.11: 86264096-86264096 on Build GRCh38
rs373200988 Chr.11: 86264298-86264298 on Build GRCh38

View Full Product Details